Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03464630_pri
          Assay Name:
          rno-mir-96
          Stem-loop ID:
          rno-mir-183 rno-mir-3553 rno-mir-96
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.4: 57463569 - 57463678 [-] on Build Rnor_6.0
          Stem-loop Location:
          Chr.4: 57463569 - 57463678 [-] on Build Rnor_6.0 Chr.4: 57463572 - 57463677 [+] on Build Rnor_6.0 Chr.4: 57463366 - 57463471 [-] on Build Rnor_6.0
          Mature miRNA ID:
          rno-miR-183-3p rno-miR-183-5p rno-miR-3553 rno-miR-96-5p rno-miR-96-3p
          Species:
          Rat
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Rn03464630_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          71

          Chromosome Location:

          Chr. 4 - 57463224 - 57463224 [+] on 57463224 Build Rnor_6.0

          Species:

          Rat

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-183-3p MIMAT0017141
          Rat rno-miR-183-5p MIMAT0000860
          Rat rno-miR-3553 MIMAT0017814
          Rat rno-miR-96-5p MIMAT0000818
          Rat rno-miR-96-3p MIMAT0017110

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-183
          Stem-loop Accession # MI0000928
          Stem-loop Sequence
          CCAGAGAGUGUGACUCCUGUCCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCGCGA
          Chromosome Location Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017141
          miRBase ID: rno-miR-183-3p
          Mature miRNA Sequence: UGAAUUACCGAAGGGCCAUAA
          Chromosome Location: Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0

          miRBase Accession #: MIMAT0000860
          miRBase ID: rno-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0

          Gene Family ID MIPF0000066, mir-183
          Distance to Amplicon 305 bases
          Stem-loop ID rno-mir-3553
          Stem-loop Accession # MI0015410
          Stem-loop Sequence
          CGGAUCUGUCUCUGCUCUGUUUAUGGCCCUUCGGUAAUUCACUGACCGAGACUGUUCACAGUGAAUUCUACCAGUGCCAUACACAGGACAGGAGUCACACUCUCUG
          Chromosome Location Chr. 4 - 57463572 - 57463677 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017814
          miRBase ID: rno-miR-3553
          Mature miRNA Sequence: CAGUGAAUUCUACCAGUGCCAUA
          Chromosome Location: Chr. 4 - 57463572 - 57463677 [+] on Build Rnor_6.0

          Gene Family ID
          Distance to Amplicon 308 bases
          Stem-loop ID rno-mir-96
          Stem-loop Accession # MI0000881
          Stem-loop Sequence
          CCAGUACCAUCUGCUUGGCCGAUUUUGGCACUAGCACAUUUUUGCUUGUGUCUCUCCGCUCUGAGCAAUCAUGUGCAGUGCCAAUAUGGGAAAAGCGGGCUGCUGC
          Chromosome Location Chr. 4 - 57463366 - 57463471 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000818
          miRBase ID: rno-miR-96-5p
          Mature miRNA Sequence: UUUGGCACUAGCACAUUUUUGCU
          Chromosome Location: Chr. 4 - 57463366 - 57463471 [-] on Build Rnor_6.0

          miRBase Accession #: MIMAT0017110
          miRBase ID: rno-miR-96-3p
          Mature miRNA Sequence: CAAUCAUGUGCAGUGCCAAUAU
          Chromosome Location: Chr. 4 - 57463366 - 57463471 [-] on Build Rnor_6.0

          Gene Family ID MIPF0000072, mir-96
          Distance to Amplicon 102 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10422
          Ambion® Pre-miR™ miRNA Precursor : PM10422
          TaqMan™ MicroRNA Assay : 000186
          mirVana® miRNA inhibitor : MH10422
          mirVana® miRNA mimic : MC10422
          TaqMan™ Advanced miRNA Assay : rno481282_mir
          TaqMan™ Advanced miRNA Assay : 478215_mir
          TaqMan™ Advanced miRNA Assay : mmu481282_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20229
          Ambion® Pre-miR™ miRNA Precursor : PM20229
          TaqMan™ MicroRNA Assay : 462586_mat
          mirVana® miRNA inhibitor : MH20229
          mirVana® miRNA mimic : MC20229
          TaqMan™ Advanced miRNA Assay : rno481096_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19308
          Ambion® Pre-miR™ miRNA Precursor : PM19308
          TaqMan™ MicroRNA Assay : 464122_mat
          mirVana® miRNA inhibitor : MH19308
          mirVana® miRNA mimic : MC19308
          TaqMan™ Advanced miRNA Assay : rno481281_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19447
          Ambion® Pre-miR™ miRNA Precursor : PM19447
          TaqMan™ MicroRNA Assay : 463953_mat
          mirVana® miRNA inhibitor : MH19447
          mirVana® miRNA mimic : MC19447
          TaqMan™ Advanced miRNA Assay : rno481329_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12830
          Ambion® Pre-miR™ miRNA Precursor : PM12830
          TaqMan™ MicroRNA Assay : 002269
          mirVana® miRNA inhibitor : MH12830
          mirVana® miRNA mimic : MC12830
          TaqMan™ Advanced miRNA Assay : 477937_mir
          TaqMan™ Advanced miRNA Assay : rno481330_mir
          TaqMan™ Advanced miRNA Assay : mmu482690_mir


          miRBase Alias:

          Rat: rno-mir-183 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline