Search
Search

| Assay Name: | hsa-miR-892c-3p |
|---|---|
| miRBase Accession Number: | MI0022560 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CACUGUUUCCUUUCUGAGUGGA |
| Species: | Human, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | CACUGUUUCCUUUCUGAGUGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-892c-3p | MIMAT0025858 |
| Rhesus monkey | mml-miR-892c-3p | MIMAT0028385 |