Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03295718_pri
          Assay Name:
          hsa-mir-888
          Stem-loop ID:
          hsa-mir-888 hsa-mir-890 hsa-mir-892c
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.X: 145994784 - 145994860 [-] on Build GRCh38
          Stem-loop Location:
          Chr.X: 145994784 - 145994860 [-] on Build GRCh38 Chr.X: 145994275 - 145994351 [-] on Build GRCh38 Chr.X: 145992750 - 145992826 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-888-3p hsa-miR-888-5p hsa-miR-890 hsa-miR-892c-3p hsa-miR-892c-5p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03295718_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          92

          Chromosome Location:

          Chr. X - 145994579 - 145994579 [-] on 145994579 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-888-3p MIMAT0004917
          Human hsa-miR-888-5p MIMAT0004916
          Human hsa-miR-890 MIMAT0004912
          Human hsa-miR-892c-3p MIMAT0025858
          Human hsa-miR-892c-5p MIMAT0025857

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-888
          Stem-loop Accession # MI0005537
          Stem-loop Sequence
          GGCAGUGCUCUACUCAAAAAGCUGUCAGUCACUUAGAUUACAUGUGACUGACACCUCUUUGGGUGAAGGAAGGCUCA
          Chromosome Location Chr. X - 145994784 - 145994860 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004917
          miRBase ID: hsa-miR-888-3p
          Mature miRNA Sequence: GACUGACACCUCUUUGGGUGAA
          Chromosome Location: Chr. X - 145994784 - 145994860 [-] on Build GRCh38

          miRBase Accession #: MIMAT0004916
          miRBase ID: hsa-miR-888-5p
          Mature miRNA Sequence: UACUCAAAAAGCUGUCAGUCA
          Chromosome Location: Chr. X - 145994784 - 145994860 [-] on Build GRCh38

          Gene Family ID MIPF0000386, mir-743
          Distance to Amplicon 145 bases
          Stem-loop ID hsa-mir-890
          Stem-loop Accession # MI0005533
          Stem-loop Sequence
          GGAAGUGCCCUACUUGGAAAGGCAUCAGUUGCUUAGAUUACAUGUAACUAUUCCCUUUCUGAGUAGAGUAAGUCUUA
          Chromosome Location Chr. X - 145994275 - 145994351 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004912
          miRBase ID: hsa-miR-890
          Mature miRNA Sequence: UACUUGGAAAGGCAUCAGUUG
          Chromosome Location: Chr. X - 145994275 - 145994351 [-] on Build GRCh38

          Gene Family ID MIPF0000386, mir-743
          Distance to Amplicon 197 bases
          Stem-loop ID hsa-mir-892c
          Stem-loop Accession # MI0022560
          Stem-loop Sequence
          GGCAGUGCCCUAUUCAGAAAGGUGCCAGUCACUUAGAUUACAUGUCACUGUUUCCUUUCUGAGUGGAGUAGGGCUUA
          Chromosome Location Chr. X - 145992750 - 145992826 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0025858
          miRBase ID: hsa-miR-892c-3p
          Mature miRNA Sequence: CACUGUUUCCUUUCUGAGUGGA
          Chromosome Location: Chr. X - 145992750 - 145992826 [-] on Build GRCh38

          miRBase Accession #: MIMAT0025857
          miRBase ID: hsa-miR-892c-5p
          Mature miRNA Sequence: UAUUCAGAAAGGUGCCAGUCA
          Chromosome Location: Chr. X - 145992750 - 145992826 [-] on Build GRCh38

          Gene Family ID MIPF0000386, mir-743
          Distance to Amplicon 1722 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM24244
          Ambion® Pre-miR™ miRNA Precursor : PM24244
          TaqMan™ MicroRNA Assay : 477431_mat
          mirVana® miRNA inhibitor : MH24244
          mirVana® miRNA mimic : MC24244
          TaqMan™ Advanced miRNA Assay : 479199_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM25473
          Ambion® Pre-miR™ miRNA Precursor : PM25473
          TaqMan™ MicroRNA Assay : 470966_mat
          mirVana® miRNA inhibitor : MH25473
          mirVana® miRNA mimic : MC25473
          TaqMan™ Advanced miRNA Assay : 479200_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12760
          Ambion® Pre-miR™ miRNA Precursor : PM12760
          TaqMan™ MicroRNA Assay : 002213
          mirVana® miRNA inhibitor : MH12760
          mirVana® miRNA mimic : MC12760
          TaqMan™ Advanced miRNA Assay : 479191_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12400
          Ambion® Pre-miR™ miRNA Precursor : PM12400
          TaqMan™ MicroRNA Assay : 002212
          mirVana® miRNA inhibitor : MH12400
          mirVana® miRNA mimic : MC12400
          TaqMan™ Advanced miRNA Assay : 479192_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12802
          Ambion® Pre-miR™ miRNA Precursor : PM12802
          TaqMan™ MicroRNA Assay : 002209
          mirVana® miRNA inhibitor : MH12802
          mirVana® miRNA mimic : MC12802
          TaqMan™ Advanced miRNA Assay : 479194_mir


          miRBase Alias:

          Human: hsa-mir-890 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline