Search
Search

| Assay Name: | hsa-miR-890 |
|---|---|
| miRBase Accession Number: | MI0005533 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUUGGAAAGGCAUCAGUUG |
| Species: | Human, Pan troglodytes, Pongo pygmaeus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UACUUGGAAAGGCAUCAGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-890 | MIMAT0004912 |
| Pan troglodytes | ptr-miR-890 | MIMAT0008332 |
| Pongo pygmaeus | ppy-miR-890 | MIMAT0016125 |