Search
Search

| Assay Name: | hsa-miR-412-5p |
|---|---|
| Stem-loop Accession Number: | MI0002464 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUCGACCAGUUGGAAAGUAAU |
| Species: | Human, Dasypus novemcinctus, Eptesicus fuscus, Goat, Guinea pig, Pan paniscus, Sheep |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000192, mir-412 |
Mature miRNA Sequence: | UGGUCGACCAGUUGGAAAGUAAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-412-5p | MIMAT0026557 |
| Dasypus novemcinctus | dno-miR-412-5p | MIMAT0047876 |
| Eptesicus fuscus | efu-miR-412 | MIMAT0035043 |
| Goat | chi-miR-412-5p | MIMAT0036217 |
| Guinea pig | cpo-miR-412-5p | MIMAT0047239 |
| Pan paniscus | ppa-miR-412 | MIMAT0049035 |
| View More Rows | ||
| Sheep | oar-miR-412-3p | MIMAT0019329 |