Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC21335
          Assay Name: hsa-miR-412-5p
          miRBase Accession Number: MI0002464
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065447 - 101065537 [+] on Build GRCh38
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Species: Human, Dasypus novemcinctus, Eptesicus fuscus, Goat, Guinea pig, Pan paniscus, Sheep
          Product Type: mirVana® miRNA mimic
          Assay ID MC21335

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000192, mir-412

          Mature miRNA Sequence:

          UGGUCGACCAGUUGGAAAGUAAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-412-5p MIMAT0026557
          Dasypus novemcinctus dno-miR-412-5p MIMAT0047876
          Eptesicus fuscus efu-miR-412 MIMAT0035043
          Goat chi-miR-412-5p MIMAT0036217
          Guinea pig cpo-miR-412-5p MIMAT0047239
          Pan paniscus ppa-miR-412 MIMAT0049035
          View More Rows
          Sheep oar-miR-412-3p MIMAT0019329

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-412
          Stem-loop Accession # MI0002464
          Stem-loop Sequence
          CUGGGGUACGGGGAUGGAUGGUCGACCAGUUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGCCGUCCGUAUCCGCUGCAG
          Chromosome Location Chr. 14 - 101065447 - 101065537 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026557
          miRBase ID: hsa-miR-412-5p
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: Chr. 14 - 101065447 - 101065537 [+] on Build GRCh38

          Dasypus novemcinctus

          Stem-loop ID dno-mir-412
          Stem-loop Accession # MI0039103
          Stem-loop Sequence
          UGGUCGACCAGUUGGAAAGUAAUUGUUCAGAAUGUACUUCACCUGGUCCACUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047876
          miRBase ID: dno-miR-412-5p
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-412
          Stem-loop Accession # MI0028728
          Stem-loop Sequence
          ACAGGAUGGGUGGUCGACCAGUUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGCCGUCCGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035043
          miRBase ID: efu-miR-412
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-412
          Stem-loop Accession # MI0030786
          Stem-loop Sequence
          AGGGAAGAACGUCAGUACCAGCAACCACUCGGGUACAGGACGGAUGGUCGACCAGUUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGCUGUCCGUACCCACUGCAGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036217
          miRBase ID: chi-miR-412-5p
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-412
          Stem-loop Accession # MI0038768
          Stem-loop Sequence
          UGGUCGACCAGUUGGAAAGUAAUUGUCUCUAAUGUACUUCACCUGGUCCACUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047239
          miRBase ID: cpo-miR-412-5p
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-412
          Stem-loop Accession # MI0039743
          Stem-loop Sequence
          UGGUCGACCAGUUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049035
          miRBase ID: ppa-miR-412
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-412
          Stem-loop Accession # MI0016959
          Stem-loop Sequence
          AGGGAAGAACGUCAGUACCAGCAACCACUCUGGGGUACAGGACGGAUGGUCGACCAGUUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGCUGUCCGUACCCACUGCAGCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019329
          miRBase ID: oar-miR-412-3p
          Mature miRNA Sequence: UGGUCGACCAGUUGGAAAGUAAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04225933_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM21335
          Ambion® Pre-miR™ miRNA Precursor : PM21335
          TaqMan™ MicroRNA Assay : 462338_mat
          mirVana® miRNA inhibitor : MH21335
          TaqMan™ Advanced miRNA Assay : 478874_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline