Search
Search

| Assay Name: | hsa-miR-487b-5p |
|---|---|
| Stem-loop Accession Number: | MI0003530 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGGUUAUCCCUGUCCUGUUCG |
| Species: | Human, Guinea pig, Pig, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GUGGUUAUCCCUGUCCUGUUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-487b-5p | MIMAT0026614 |
| Guinea pig | cpo-miR-487b-5p | MIMAT0047281 |
| Pig | ssc-miR-487b | MIMAT0017973 |
| Pteropus alecto | pal-miR-487b-5p | MIMAT0040019 |
| Rabbit | ocu-miR-487b-5p | MIMAT0048473 |
| Rhesus monkey | mml-miR-487b-5p | MIMAT0026881 |