Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19387
          Assay Name: hsa-miR-487b-5p
          miRBase Accession Number: MI0003530
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101046455 - 101046538 [+] on Build GRCh38
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Species: Human, Guinea pig, Pig, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC19387

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          GUGGUUAUCCCUGUCCUGUUCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-487b-5p MIMAT0026614
          Guinea pig cpo-miR-487b-5p MIMAT0047281
          Pig ssc-miR-487b MIMAT0017973
          Pteropus alecto pal-miR-487b-5p MIMAT0040019
          Rabbit ocu-miR-487b-5p MIMAT0048473
          Rhesus monkey mml-miR-487b-5p MIMAT0026881

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-487b
          Stem-loop Accession # MI0003530
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location Chr. 14 - 101046455 - 101046538 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026614
          miRBase ID: hsa-miR-487b-5p
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Chromosome Location: Chr. 14 - 101046455 - 101046538 [+] on Build GRCh38

          Guinea pig

          Stem-loop ID cpo-mir-487b
          Stem-loop Accession # MI0038790
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUCACUCACGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047281
          miRBase ID: cpo-miR-487b-5p
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-487b
          Stem-loop Accession # MI0015928
          Stem-loop Sequence
          UACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017973
          miRBase ID: ssc-miR-487b
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-487b
          Stem-loop Accession # MI0032451
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040019
          miRBase ID: pal-miR-487b-5p
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-487b
          Stem-loop Accession # MI0039419
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUUAUUCAUGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048473
          miRBase ID: ocu-miR-487b-5p
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-487b
          Stem-loop Accession # MI0007761
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCGUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026881
          miRBase ID: mml-miR-487b-5p
          Mature miRNA Sequence: GUGGUUAUCCCUGUCCUGUUCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303552_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19387
          Ambion® Pre-miR™ miRNA Precursor : PM19387
          TaqMan™ MicroRNA Assay : 463617_mat
          mirVana® miRNA inhibitor : MH19387
          TaqMan™ Advanced miRNA Assay : 478938_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline