Search
Search

| Assay Name: | hsa-miR-186-3p |
|---|---|
| Stem-loop Accession Number: | MI0000483 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCCCAAAGGUGAAUUUUUUGGG |
| Species: | Human, Dasypus novemcinctus, Guinea pig, Pig, Rabbit |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | GCCCAAAGGUGAAUUUUUUGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-186-3p | MIMAT0004612 |
| Dasypus novemcinctus | dno-miR-186-3p | MIMAT0047708 |
| Guinea pig | cpo-miR-186-3p | MIMAT0047057 |
| Pig | ssc-miR-186-3p | MIMAT0037321 |
| Rabbit | ocu-miR-186-3p | MIMAT0048294 |