Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03303293_pri
Assay Name:
hsa-mir-186
Stem-loop ID:
hsa-mir-186
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.1: 71067631 - 71067716 [-] on Build GRCh38
Stem-loop Location:
Chr.1: 71067631 - 71067716 [-] on Build GRCh38
Mature miRNA ID:
hsa-miR-186-5p
hsa-miR-186-3p
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03303293_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
169
Chromosome Location:
Chr. 1 - 71067332 - 71067332 [-] on 71067332 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-186-5p
MIMAT0000456
Human
hsa-miR-186-3p
MIMAT0004612
Stem-loop Details
Human
Stem-loop ID
hsa-mir-186
Stem-loop Accession #
MI0000483
Stem-loop Sequence
UGCUUGUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGCU
Chromosome Location
Chr. 1 - 71067631 - 71067716 [-] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0000456
miRBase ID: hsa-miR-186-5p
Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
Chromosome Location: Chr. 1 - 71067631 - 71067716 [-] on Build GRCh38
miRBase Accession #: MIMAT0004612
miRBase ID: hsa-miR-186-3p
Mature miRNA Sequence: GCCCAAAGGUGAAUUUUUUGGG
Chromosome Location: Chr. 1 - 71067631 - 71067716 [-] on Build GRCh38
Gene Family ID
MIPF0000109,
mir-186
Distance to Amplicon
204 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM11753
Ambion
®
Pre-miR
™
miRNA Precursor :
PM11753
TaqMan
™
MicroRNA Assay :
002285
mir
Vana
®
miRNA inhibitor :
MH11753
mir
Vana
®
miRNA mimic :
MC11753
TaqMan
™
Advanced miRNA Assay :
mmu480966_mir
TaqMan
™
Advanced miRNA Assay :
477940_mir
TaqMan
™
Advanced miRNA Assay :
rno480966_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12359
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12359
TaqMan
™
MicroRNA Assay :
002105
mir
Vana
®
miRNA inhibitor :
MH12359
mir
Vana
®
miRNA mimic :
MC12359
TaqMan
™
Advanced miRNA Assay :
479544_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline