Search
Search

| Assay Name: | hsa-miR-186 |
|---|---|
| miRBase Accession Number: | MI0000483 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAAGAAUUCUCCUUUUGGGCU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Daubentonia madagascariensis, Dog, Goat, Horse, Nomascus leucogenys, Ornithorhynchus anatinus, Papio hamadryas, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | CAAAGAAUUCUCCUUUUGGGCU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-186-5p | MIMAT0000456 | |
| Mouse | mmu-miR-186-5p | MIMAT0000215 | |
| Rat | rno-miR-186-5p | MIMAT0000863 | |
| Bovine | bta-miR-186 | MIMAT0003818 | |
| Callithrix jacchus | cja-miR-186 | MIMAT0039388 | |
| Daubentonia madagascariensis | dma-miR-186 | MIMAT0049271 | |
| View More Rows | |||
| Dog | cfa-miR-186 | MIMAT0006694 | |
| Goat | chi-miR-186-5p | MIMAT0036010 | |
| Horse | eca-miR-186 | MIMAT0012956 | |
| Nomascus leucogenys | nle-miR-186 | MIMAT0049448 | |
| Ornithorhynchus anatinus | oan-miR-186-5p | MIMAT0006931 | |
| Papio hamadryas | pha-miR-186 | MIMAT0049515 | |
| Pongo pygmaeus | ppy-miR-186 | MIMAT0015782 | |
| Rhesus monkey | mml-miR-186-5p | MIMAT0006220 | |
| Tupaia chinensis | tch-miR-186-5p | MIMAT0036622 | |