Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480966_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-186-5p
          Stem-loop Accession Number: MI0000483
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 71067631 - 71067716 [-] on Build GRCh38
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Daubentonia madagascariensis, Dog, Goat, Horse, Nomascus leucogenys, Ornithorhynchus anatinus, Papio hamadryas, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480966_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000109, mir-186

          Mature miRNA Sequence:

          CAAAGAAUUCUCCUUUUGGGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-186-5p MIMAT0000456
          Mouse mmu-miR-186-5p MIMAT0000215
          Rat rno-miR-186-5p MIMAT0000863
          Bovine bta-miR-186 MIMAT0003818
          Callithrix jacchus cja-miR-186 MIMAT0039388
          Daubentonia madagascariensis dma-miR-186 MIMAT0049271
          View More Rows
          Dog cfa-miR-186 MIMAT0006694
          Goat chi-miR-186-5p MIMAT0036010
          Horse eca-miR-186 MIMAT0012956
          Nomascus leucogenys nle-miR-186 MIMAT0049448
          Ornithorhynchus anatinus oan-miR-186-5p MIMAT0006931
          Papio hamadryas pha-miR-186 MIMAT0049515
          Pongo pygmaeus ppy-miR-186 MIMAT0015782
          Rhesus monkey mml-miR-186-5p MIMAT0006220
          Tupaia chinensis tch-miR-186-5p MIMAT0036622

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-186
          Stem-loop Accession # MI0000483
          Stem-loop Sequence
          UGCUUGUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGCU
          Chromosome Location Chr. 1 - 71067631 - 71067716 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000456
          miRBase ID: hsa-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: Chr. 1 - 71067631 - 71067716 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-186
          Stem-loop Accession # MI0000228
          Stem-loop Sequence
          ACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUCAUUUUAUUUUAAGCCCUAAGGUGAAUUUUUUGGGAAGU
          Chromosome Location Chr. 3 - 157544279 - 157544349 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000215
          miRBase ID: mmu-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: Chr. 3 - 157544279 - 157544349 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-186
          Stem-loop Accession # MI0000931
          Stem-loop Sequence
          UGCUUACAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUCAUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGCU
          Chromosome Location Chr. 2 - 263873759 - 263873844 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000863
          miRBase ID: rno-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: Chr. 2 - 263873759 - 263873844 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-186
          Stem-loop Accession # MI0005033
          Stem-loop Sequence
          UGCUUAUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGAUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003818
          miRBase ID: bta-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-186
          Stem-loop Accession # MI0031907
          Stem-loop Sequence
          GCUUGUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039388
          miRBase ID: cja-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-186
          Stem-loop Accession # MI0040000
          Stem-loop Sequence
          CAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAACCCAAAGGUGAAUUUUUUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049271
          miRBase ID: dma-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-186
          Stem-loop Accession # MI0008108
          Stem-loop Sequence
          CAAAGAAUUCUCCUUUUGGGCUUUCUGAUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006694
          miRBase ID: cfa-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-186
          Stem-loop Accession # MI0030657
          Stem-loop Sequence
          UGCUUAUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGAUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036010
          miRBase ID: chi-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-186
          Stem-loop Accession # MI0012708
          Stem-loop Sequence
          ACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGAUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012956
          miRBase ID: eca-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-186
          Stem-loop Accession # MI0040196
          Stem-loop Sequence
          CAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049448
          miRBase ID: nle-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-186
          Stem-loop Accession # MI0006741
          Stem-loop Sequence
          GCUCAUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUAUGCUUGGUCACAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006931
          miRBase ID: oan-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-186
          Stem-loop Accession # MI0040280
          Stem-loop Sequence
          CAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUCUUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049515
          miRBase ID: pha-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-186
          Stem-loop Accession # MI0014850
          Stem-loop Sequence
          UGCUUGUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUUUUGGGAAGUUUGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015782
          miRBase ID: ppy-miR-186
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-186
          Stem-loop Accession # MI0007650
          Stem-loop Sequence
          UGCUUGUAACUUUCCAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUUAUUUUAAGCCCAAAGGUGAAUUUCUUGGGAAGUUUGAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006220
          miRBase ID: mml-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-186
          Stem-loop Accession # MI0031295
          Stem-loop Sequence
          CAAAGAAUUCUCCUUUUGGGCUUUCUGGUUUCGUUUUAAGCCCAAAGGUGAAUUUUUUGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036622
          miRBase ID: tch-miR-186-5p
          Mature miRNA Sequence: CAAAGAAUUCUCCUUUUGGGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303293_pri
          TaqMan™ Pri-miRNA Assay : Mm03306448_pri
          TaqMan™ Pri-miRNA Assay : Rn03464637_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11753
          Ambion® Pre-miR™ miRNA Precursor : PM11753
          TaqMan™ MicroRNA Assay : 002285
          mirVana® miRNA inhibitor : MH11753
          mirVana® miRNA mimic : MC11753
          TaqMan™ Advanced miRNA Assay : 477940_mir
          TaqMan™ Advanced miRNA Assay : rno480966_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline