Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10034
          Assay Name: hsa-miR-328-3p
          miRBase Accession Number: MI0000804
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 67202321 - 67202395 [-] on Build GRCh38
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Horse, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Tupaia chinensis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10034
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000203, mir-328

          Mature miRNA Sequence:

          CUGGCCCUCUCUGCCCUUCCGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-328-3p MIMAT0000752
          Mouse mmu-miR-328-3p MIMAT0000565
          Rat rno-miR-328a-3p MIMAT0000564
          Bovine bta-miR-328 MIMAT0009287
          Callithrix jacchus cja-miR-328 MIMAT0039405
          Cricetulus griseus cgr-miR-328 MIMAT0023914
          View More Rows
          Dog cfa-miR-328 MIMAT0006688
          Goat chi-miR-328-3p MIMAT0036139
          Horse eca-miR-328 MIMAT0012929
          Pan paniscus ppa-miR-328 MIMAT0049093
          Pan troglodytes ptr-miR-328 MIMAT0008100
          Pig ssc-miR-328 MIMAT0013949
          Pongo pygmaeus ppy-miR-328 MIMAT0015831
          Tupaia chinensis tch-miR-328 MIMAT0036581

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-328
          Stem-loop Accession # MI0000804
          Stem-loop Sequence
          UGGAGUGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUG
          Chromosome Location Chr. 16 - 67202321 - 67202395 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000752
          miRBase ID: hsa-miR-328-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: Chr. 16 - 67202321 - 67202395 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-328
          Stem-loop Accession # MI0000603
          Stem-loop Sequence
          CUGUCUCGGAGCCUGGGGCAGGGGGGCAGGAGGGGCUCAGGGAGAAAGUAUCUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGUCCCCAAGU
          Chromosome Location Chr. 8 - 105308364 - 105308460 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000565
          miRBase ID: mmu-miR-328-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: Chr. 8 - 105308364 - 105308460 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-328a
          Stem-loop Accession # MI0000602
          Stem-loop Sequence
          UGGGGCAGGGGGGCAGGAGGGGCUCAGGGAGAAAGCAUCUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGUCCCCAAAU
          Chromosome Location Chr. 19 - 37263199 - 37263282 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000564
          miRBase ID: rno-miR-328a-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: Chr. 19 - 37263199 - 37263282 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-328
          Stem-loop Accession # MI0009800
          Stem-loop Sequence
          CUGUUUCGGAGCCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAUAGUGUGUGCUGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009287
          miRBase ID: bta-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-328
          Stem-loop Accession # MI0031924
          Stem-loop Sequence
          GGAGCCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUGCAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039405
          miRBase ID: cja-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-328
          Stem-loop Accession # MI0020496
          Stem-loop Sequence
          CCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGUCUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023914
          miRBase ID: cgr-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-328
          Stem-loop Accession # MI0008099
          Stem-loop Sequence
          AGGGGGGCAGGAAGGGCUCAGGGAGAAAGUGUGUGCAGCCCCUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006688
          miRBase ID: cfa-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-328
          Stem-loop Accession # MI0030742
          Stem-loop Sequence
          CUGUUUCGGAGCCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAUAGUGUGUGCUGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCCGUUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036139
          miRBase ID: chi-miR-328-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-328
          Stem-loop Accession # MI0012684
          Stem-loop Sequence
          GGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGUGUGCAGCCCCUGGCCCUCUCUGCCCUUCCGUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012929
          miRBase ID: eca-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-328
          Stem-loop Accession # MI0039803
          Stem-loop Sequence
          GGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUACAGCCCCUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049093
          miRBase ID: ppa-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-328
          Stem-loop Accession # MI0008618
          Stem-loop Sequence
          UGGAGUGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008100
          miRBase ID: ptr-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-328
          Stem-loop Accession # MI0013158
          Stem-loop Sequence
          GGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGUGUGCCGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCCGUUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013949
          miRBase ID: ssc-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-328
          Stem-loop Accession # MI0014895
          Stem-loop Sequence
          UGGAGUGGGGGGGCAGGAGGAGCUCAGGGAGAAAGUGCAUGCAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015831
          miRBase ID: ppy-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-328
          Stem-loop Accession # MI0031245
          Stem-loop Sequence
          GGGGGGCAGGAGGGGCUCAGGGAGGAAGCCUGUAUAGCCCCUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036581
          miRBase ID: tch-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465910_pri
          TaqMan™ Pri-miRNA Assay : Mm03306951_pri
          TaqMan™ Pri-miRNA Assay : Hs03303583_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10034
          TaqMan™ MicroRNA Assay : 000543
          mirVana® miRNA inhibitor : MH10034
          mirVana® miRNA mimic : MC10034
          TaqMan™ Advanced miRNA Assay : 478028_mir
          TaqMan™ Advanced miRNA Assay : mmu481059_mir
          TaqMan™ Advanced miRNA Assay : rno481059_mir


          miRBase Alias:

          Human: hsa-miR-328 (v19)
          Mouse: mmu-miR-328 (v17)
          Rat: rno-miR-328a (v18)
          Rat: rno-miR-328 (v15)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline