Search
Search

| Assay Name: | mmu-miR-328-3p |
|---|---|
| Stem-loop Accession Number: | MI0000804 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGGCCCUCUCUGCCCUUCCGU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Horse, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000203, mir-328 |
Mature miRNA Sequence: | CUGGCCCUCUCUGCCCUUCCGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-328-3p | MIMAT0000752 |
| Mouse | mmu-miR-328-3p | MIMAT0000565 |
| Rat | rno-miR-328a-3p | MIMAT0000564 |
| Bovine | bta-miR-328 | MIMAT0009287 |
| Callithrix jacchus | cja-miR-328 | MIMAT0039405 |
| Cricetulus griseus | cgr-miR-328 | MIMAT0023914 |
| View More Rows | ||
| Dog | cfa-miR-328 | MIMAT0006688 |
| Goat | chi-miR-328-3p | MIMAT0036139 |
| Horse | eca-miR-328 | MIMAT0012929 |
| Pan paniscus | ppa-miR-328 | MIMAT0049093 |
| Pan troglodytes | ptr-miR-328 | MIMAT0008100 |
| Pig | ssc-miR-328 | MIMAT0013949 |
| Pongo pygmaeus | ppy-miR-328 | MIMAT0015831 |
| Tupaia chinensis | tch-miR-328 | MIMAT0036581 |