Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478028_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-328-3p
          Stem-loop Accession Number: MI0000804
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 67202321 - 67202395 [-] on Build GRCh38
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Horse, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478028_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000203, mir-328

          Mature miRNA Sequence:

          CUGGCCCUCUCUGCCCUUCCGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-328-3p MIMAT0000752
          Mouse mmu-miR-328-3p MIMAT0000565
          Rat rno-miR-328a-3p MIMAT0000564
          Bovine bta-miR-328 MIMAT0009287
          Callithrix jacchus cja-miR-328 MIMAT0039405
          Cricetulus griseus cgr-miR-328 MIMAT0023914
          View More Rows
          Dog cfa-miR-328 MIMAT0006688
          Goat chi-miR-328-3p MIMAT0036139
          Horse eca-miR-328 MIMAT0012929
          Pan paniscus ppa-miR-328 MIMAT0049093
          Pan troglodytes ptr-miR-328 MIMAT0008100
          Pig ssc-miR-328 MIMAT0013949
          Pongo pygmaeus ppy-miR-328 MIMAT0015831
          Tupaia chinensis tch-miR-328 MIMAT0036581

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-328
          Stem-loop Accession # MI0000804
          Stem-loop Sequence
          UGGAGUGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUG
          Chromosome Location Chr. 16 - 67202321 - 67202395 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000752
          miRBase ID: hsa-miR-328-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: Chr. 16 - 67202321 - 67202395 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-328
          Stem-loop Accession # MI0000603
          Stem-loop Sequence
          CUGUCUCGGAGCCUGGGGCAGGGGGGCAGGAGGGGCUCAGGGAGAAAGUAUCUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGUCCCCAAGU
          Chromosome Location Chr. 8 - 105308364 - 105308460 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000565
          miRBase ID: mmu-miR-328-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: Chr. 8 - 105308364 - 105308460 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-328a
          Stem-loop Accession # MI0000602
          Stem-loop Sequence
          UGGGGCAGGGGGGCAGGAGGGGCUCAGGGAGAAAGCAUCUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGUCCCCAAAU
          Chromosome Location Chr. 19 - 37263199 - 37263282 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000564
          miRBase ID: rno-miR-328a-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: Chr. 19 - 37263199 - 37263282 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-328
          Stem-loop Accession # MI0009800
          Stem-loop Sequence
          CUGUUUCGGAGCCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAUAGUGUGUGCUGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009287
          miRBase ID: bta-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-328
          Stem-loop Accession # MI0031924
          Stem-loop Sequence
          GGAGCCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUGCAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039405
          miRBase ID: cja-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-328
          Stem-loop Accession # MI0020496
          Stem-loop Sequence
          CCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGUCUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023914
          miRBase ID: cgr-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-328
          Stem-loop Accession # MI0008099
          Stem-loop Sequence
          AGGGGGGCAGGAAGGGCUCAGGGAGAAAGUGUGUGCAGCCCCUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006688
          miRBase ID: cfa-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-328
          Stem-loop Accession # MI0030742
          Stem-loop Sequence
          CUGUUUCGGAGCCUGGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAUAGUGUGUGCUGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCCGUUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036139
          miRBase ID: chi-miR-328-3p
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-328
          Stem-loop Accession # MI0012684
          Stem-loop Sequence
          GGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGUGUGCAGCCCCUGGCCCUCUCUGCCCUUCCGUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012929
          miRBase ID: eca-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-328
          Stem-loop Accession # MI0039803
          Stem-loop Sequence
          GGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUACAGCCCCUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049093
          miRBase ID: ppa-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-328
          Stem-loop Accession # MI0008618
          Stem-loop Sequence
          UGGAGUGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGCAUACAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008100
          miRBase ID: ptr-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-328
          Stem-loop Accession # MI0013158
          Stem-loop Sequence
          GGAGCGGGGGGGCAGGAGGGGCUCAGGGAGAAAGUGUGUGCCGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCCGUUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013949
          miRBase ID: ssc-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-328
          Stem-loop Accession # MI0014895
          Stem-loop Sequence
          UGGAGUGGGGGGGCAGGAGGAGCUCAGGGAGAAAGUGCAUGCAGCCCCUGGCCCUCUCUGCCCUUCCGUCCCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015831
          miRBase ID: ppy-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-328
          Stem-loop Accession # MI0031245
          Stem-loop Sequence
          GGGGGGCAGGAGGGGCUCAGGGAGGAAGCCUGUAUAGCCCCUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036581
          miRBase ID: tch-miR-328
          Mature miRNA Sequence: CUGGCCCUCUCUGCCCUUCCGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465910_pri
          TaqMan™ Pri-miRNA Assay : Mm03306951_pri
          TaqMan™ Pri-miRNA Assay : Hs03303583_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10034
          Ambion® Pre-miR™ miRNA Precursor : PM10034
          TaqMan™ MicroRNA Assay : 000543
          mirVana® miRNA inhibitor : MH10034
          mirVana® miRNA mimic : MC10034
          TaqMan™ Advanced miRNA Assay : mmu481059_mir
          TaqMan™ Advanced miRNA Assay : rno481059_mir
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline