Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10102
          Assay Name: hsa-miR-92b-3p
          miRBase Accession Number: MI0003560
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 155195177 - 155195272 [+] on Build GRCh38
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Species: Human, Mouse, Rat, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Eptesicus fuscus, Guinea pig, Horse, Ictalurus punctatus, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Oreochromis niloticus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Salmo salar, Zebrafish
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10102
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000013, mir-25

          Mature miRNA Sequence:

          UAUUGCACUCGUCCCGGCCUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-92b-3p MIMAT0003218
          Mouse mmu-miR-92b-3p MIMAT0004899
          Rat rno-miR-92b-3p MIMAT0005340
          Astatotilapia burtoni abu-miR-92b MIMAT0042106
          Bovine bta-miR-92b MIMAT0009384
          Callithrix jacchus cja-miR-92b MIMAT0039453
          View More Rows
          Chrysemys picta cpi-miR-92b-3p MIMAT0037702
          Cricetulus griseus cgr-miR-92b-3p MIMAT0024012
          Cyprinus carpio ccr-miR-92b MIMAT0026324
          Dasypus novemcinctus dno-miR-92b-3p MIMAT0047585
          Dog cfa-miR-92b MIMAT0006703
          Eptesicus fuscus efu-miR-92b MIMAT0035160
          Guinea pig cpo-miR-92b-3p MIMAT0046932
          Horse eca-miR-92b MIMAT0012966
          Ictalurus punctatus ipu-miR-92b MIMAT0029530
          Metriaclima zebra mze-miR-92b MIMAT0042305
          Monodelphis domestica mdo-miR-92b-3p MIMAT0028519
          Neolamprologus brichardi nbr-miR-92b MIMAT0042497
          Nomascus leucogenys nle-miR-92b MIMAT0049451
          Oreochromis niloticus oni-miR-92b MIMAT0042694
          Pan paniscus ppa-miR-92b MIMAT0049080
          Pan troglodytes ptr-miR-92b MIMAT0024028
          Pig ssc-miR-92b-3p MIMAT0013909
          Pongo pygmaeus ppy-miR-92b MIMAT0015744
          Pteropus alecto pal-miR-92b-3p MIMAT0039908
          Rhesus monkey mml-miR-92b-3p MIMAT0006177
          Salmo salar ssa-miR-92b-3p MIMAT0032712
          Zebrafish dre-miR-92b-3p MIMAT0001809

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-92b
          Stem-loop Accession # MI0003560
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCC
          Chromosome Location Chr. 1 - 155195177 - 155195272 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003218
          miRBase ID: hsa-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: Chr. 1 - 155195177 - 155195272 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-92b
          Stem-loop Accession # MI0005521
          Stem-loop Sequence
          GGUGGGCGGGAGGGACGGGACGUGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCG
          Chromosome Location Chr. 3 - 89227116 - 89227198 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004899
          miRBase ID: mmu-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: Chr. 3 - 89227116 - 89227198 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-92b
          Stem-loop Accession # MI0006167
          Stem-loop Sequence
          GGUGGGCAGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCG
          Chromosome Location Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005340
          miRBase ID: rno-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-92b
          Stem-loop Accession # MI0033809
          Stem-loop Sequence
          CAGGUCCGGGAUGUGGUGCGCUGUUGUCAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042106
          miRBase ID: abu-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-92b
          Stem-loop Accession # MI0009906
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009384
          miRBase ID: bta-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-92b
          Stem-loop Accession # MI0031976
          Stem-loop Sequence
          GGGAUCCCGGGAUCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCUCCCCCUACCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039453
          miRBase ID: cja-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-92b
          Stem-loop Accession # MI0029439
          Stem-loop Sequence
          UCCAGGGCAGCAGGAGGGACGGGAUGCUGUGCAGUGUUGUUCUAUCACCAACCAAUAUUGCACUCGUCCCGGCCUCCUGCCUCCUUGGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037702
          miRBase ID: cpi-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-92b
          Stem-loop Accession # MI0020561
          Stem-loop Sequence
          GGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024012
          miRBase ID: cgr-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-92b
          Stem-loop Accession # MI0023420
          Stem-loop Sequence
          GUGUGUUUGUAGUCCCGCGGUCUGGCAGGUGUGGGAUGUUGUGCACUGUUGUUCAAUCUCCCGCCAAUAUUGCACUCGUCCCGGCCUCCCUGACCACGAGGAACACGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026324
          miRBase ID: ccr-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-92b
          Stem-loop Accession # MI0038949
          Stem-loop Sequence
          AGGGACGGGAUGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047585
          miRBase ID: dno-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-92b
          Stem-loop Accession # MI0008119
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUCUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006703
          miRBase ID: cfa-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-92b
          Stem-loop Accession # MI0028867
          Stem-loop Sequence
          GGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035160
          miRBase ID: efu-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-92b
          Stem-loop Accession # MI0038605
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCACCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046932
          miRBase ID: cpo-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-92b
          Stem-loop Accession # MI0012719
          Stem-loop Sequence
          GGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012966
          miRBase ID: eca-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-92b
          Stem-loop Accession # MI0024666
          Stem-loop Sequence
          AGGUGUGGGAUGUUGUGCAGUGUUGUUCAAUCUCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029530
          miRBase ID: ipu-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-92b
          Stem-loop Accession # MI0034065
          Stem-loop Sequence
          AGGUCCGGGAUGUGGUGCGCUGUUGUCAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042305
          miRBase ID: mze-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-92b
          Stem-loop Accession # MI0023776
          Stem-loop Sequence
          AGGGACGGGACGUGGUGCAGUGUUGUUCUUUCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028519
          miRBase ID: mdo-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-92b
          Stem-loop Accession # MI0034329
          Stem-loop Sequence
          CAGGUCCGGGACGUGGUGCGCUGUUGUCAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042497
          miRBase ID: nbr-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-92b
          Stem-loop Accession # MI0040200
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049451
          miRBase ID: nle-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-92b
          Stem-loop Accession # MI0034603
          Stem-loop Sequence
          AGGUCCGGGAUGUGGUGCACUGUUGUAAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042694
          miRBase ID: oni-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-92b
          Stem-loop Accession # MI0039788
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049080
          miRBase ID: ppa-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-92b
          Stem-loop Accession # MI0020575
          Stem-loop Sequence
          AGCGGGAUCCCGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024028
          miRBase ID: ptr-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-92b
          Stem-loop Accession # MI0013126
          Stem-loop Sequence
          GCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013909
          miRBase ID: ssc-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-92b
          Stem-loop Accession # MI0014809
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCUCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015744
          miRBase ID: ppy-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-92b
          Stem-loop Accession # MI0032394
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUUCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039908
          miRBase ID: pal-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-92b
          Stem-loop Accession # MI0007607
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006177
          miRBase ID: mml-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-92b
          Stem-loop Accession # MI0026762
          Stem-loop Sequence
          AGGUGUGGGAUGUGGUGCACUGUUGUACAAUGCCCUCCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032712
          miRBase ID: ssa-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-92b
          Stem-loop Accession # MI0001953
          Stem-loop Sequence
          UCCUACGGGCAGGGAGGUGUGGGAUGUUGUGCAGUGUUGUUCAAUCUCCCGCCAAUAUUGCACUCGUCCCGGCCUCCCUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001809
          miRBase ID: dre-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464732_pri
          TaqMan™ Pri-miRNA Assay : Hs03294072_pri
          TaqMan™ Pri-miRNA Assay : Mm03308629_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10102
          mirVana® miRNA inhibitor : MH10102
          mirVana® miRNA mimic : MC10102
          TaqMan™ Advanced miRNA Assay : rno481277_mir
          TaqMan™ Advanced miRNA Assay : mmu481277_mir
          TaqMan™ Advanced miRNA Assay : 477823_mir


          miRBase Alias:

          Human: hsa-miR-92b (v17)
          Mouse: mmu-miR-92b (v17)
          Rat: rno-miR-92b (v18)
          Danio rerio: dre-miR-92b (v20)
          Macaca mulatta: mml-miR-92b (v19)
          Staphylococcus schleiferi: ssc-miR-92b (v15)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline