Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn03464732_pri
Assay Name:
rno-mir-92b
Stem-loop ID:
rno-mir-92b
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.2: 188541034 - 188541116 [-] on Build Rnor_6.0
Stem-loop Location:
Chr.2: 188541034 - 188541116 [-] on Build Rnor_6.0
Mature miRNA ID:
rno-miR-92b-3p
rno-miR-92b-5p
Species:
Rat
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Rn03464732_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
67
Chromosome Location:
Chr. 2 - 188540881 - 188540881 [-] on 188540881 Build Rnor_6.0
Species:
Rat
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Rat
rno-miR-92b-3p
MIMAT0005340
Rat
rno-miR-92b-5p
MIMAT0017319
Stem-loop Details
Rat
Stem-loop ID
rno-mir-92b
Stem-loop Accession #
MI0006167
Stem-loop Sequence
GGUGGGCAGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCG
Chromosome Location
Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0
Mature MicroRNA
miRBase Accession #: MIMAT0005340
miRBase ID: rno-miR-92b-3p
Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
Chromosome Location: Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0
miRBase Accession #: MIMAT0017319
miRBase ID: rno-miR-92b-5p
Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
Chromosome Location: Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0
Gene Family ID
MIPF0000013,
mir-25
Distance to Amplicon
125 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10102
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10102
mir
Vana
®
miRNA inhibitor :
MH10102
mir
Vana
®
miRNA mimic :
MC10102
TaqMan
™
Advanced miRNA Assay :
rno481277_mir
TaqMan
™
Advanced miRNA Assay :
mmu481277_mir
TaqMan
™
Advanced miRNA Assay :
477823_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM20550
Ambion
®
Pre-miR
™
miRNA Precursor :
PM20550
TaqMan
™
MicroRNA Assay :
463385_mat
mir
Vana
®
miRNA inhibitor :
MH20550
mir
Vana
®
miRNA mimic :
MC20550
TaqMan
™
Advanced miRNA Assay :
rno481278_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline