Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477823_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-92b-3p
          Stem-loop Accession Number: MI0003560
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 155195177 - 155195272 [+] on Build GRCh38
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Species: Human, Mouse, Rat, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Eptesicus fuscus, Guinea pig, Horse, Ictalurus punctatus, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Oreochromis niloticus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Salmo salar, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477823_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000013, mir-25

          Mature miRNA Sequence:

          UAUUGCACUCGUCCCGGCCUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-92b-3p MIMAT0003218
          Mouse mmu-miR-92b-3p MIMAT0004899
          Rat rno-miR-92b-3p MIMAT0005340
          Astatotilapia burtoni abu-miR-92b MIMAT0042106
          Bovine bta-miR-92b MIMAT0009384
          Callithrix jacchus cja-miR-92b MIMAT0039453
          View More Rows
          Chrysemys picta cpi-miR-92b-3p MIMAT0037702
          Cricetulus griseus cgr-miR-92b-3p MIMAT0024012
          Cyprinus carpio ccr-miR-92b MIMAT0026324
          Dasypus novemcinctus dno-miR-92b-3p MIMAT0047585
          Dog cfa-miR-92b MIMAT0006703
          Eptesicus fuscus efu-miR-92b MIMAT0035160
          Guinea pig cpo-miR-92b-3p MIMAT0046932
          Horse eca-miR-92b MIMAT0012966
          Ictalurus punctatus ipu-miR-92b MIMAT0029530
          Metriaclima zebra mze-miR-92b MIMAT0042305
          Monodelphis domestica mdo-miR-92b-3p MIMAT0028519
          Neolamprologus brichardi nbr-miR-92b MIMAT0042497
          Nomascus leucogenys nle-miR-92b MIMAT0049451
          Oreochromis niloticus oni-miR-92b MIMAT0042694
          Pan paniscus ppa-miR-92b MIMAT0049080
          Pan troglodytes ptr-miR-92b MIMAT0024028
          Pig ssc-miR-92b-3p MIMAT0013909
          Pongo pygmaeus ppy-miR-92b MIMAT0015744
          Pteropus alecto pal-miR-92b-3p MIMAT0039908
          Rhesus monkey mml-miR-92b-3p MIMAT0006177
          Salmo salar ssa-miR-92b-3p MIMAT0032712
          Zebrafish dre-miR-92b-3p MIMAT0001809

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-92b
          Stem-loop Accession # MI0003560
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCC
          Chromosome Location Chr. 1 - 155195177 - 155195272 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003218
          miRBase ID: hsa-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: Chr. 1 - 155195177 - 155195272 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-92b
          Stem-loop Accession # MI0005521
          Stem-loop Sequence
          GGUGGGCGGGAGGGACGGGACGUGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCG
          Chromosome Location Chr. 3 - 89227116 - 89227198 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004899
          miRBase ID: mmu-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: Chr. 3 - 89227116 - 89227198 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-92b
          Stem-loop Accession # MI0006167
          Stem-loop Sequence
          GGUGGGCAGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCG
          Chromosome Location Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005340
          miRBase ID: rno-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-92b
          Stem-loop Accession # MI0033809
          Stem-loop Sequence
          CAGGUCCGGGAUGUGGUGCGCUGUUGUCAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042106
          miRBase ID: abu-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-92b
          Stem-loop Accession # MI0009906
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009384
          miRBase ID: bta-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-92b
          Stem-loop Accession # MI0031976
          Stem-loop Sequence
          GGGAUCCCGGGAUCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCUCCCCCUACCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039453
          miRBase ID: cja-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-92b
          Stem-loop Accession # MI0029439
          Stem-loop Sequence
          UCCAGGGCAGCAGGAGGGACGGGAUGCUGUGCAGUGUUGUUCUAUCACCAACCAAUAUUGCACUCGUCCCGGCCUCCUGCCUCCUUGGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037702
          miRBase ID: cpi-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-92b
          Stem-loop Accession # MI0020561
          Stem-loop Sequence
          GGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024012
          miRBase ID: cgr-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-92b
          Stem-loop Accession # MI0023420
          Stem-loop Sequence
          GUGUGUUUGUAGUCCCGCGGUCUGGCAGGUGUGGGAUGUUGUGCACUGUUGUUCAAUCUCCCGCCAAUAUUGCACUCGUCCCGGCCUCCCUGACCACGAGGAACACGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026324
          miRBase ID: ccr-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-92b
          Stem-loop Accession # MI0038949
          Stem-loop Sequence
          AGGGACGGGAUGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047585
          miRBase ID: dno-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-92b
          Stem-loop Accession # MI0008119
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUCUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006703
          miRBase ID: cfa-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-92b
          Stem-loop Accession # MI0028867
          Stem-loop Sequence
          GGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035160
          miRBase ID: efu-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-92b
          Stem-loop Accession # MI0038605
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCACCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046932
          miRBase ID: cpo-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-92b
          Stem-loop Accession # MI0012719
          Stem-loop Sequence
          GGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012966
          miRBase ID: eca-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-92b
          Stem-loop Accession # MI0024666
          Stem-loop Sequence
          AGGUGUGGGAUGUUGUGCAGUGUUGUUCAAUCUCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029530
          miRBase ID: ipu-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-92b
          Stem-loop Accession # MI0034065
          Stem-loop Sequence
          AGGUCCGGGAUGUGGUGCGCUGUUGUCAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042305
          miRBase ID: mze-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-92b
          Stem-loop Accession # MI0023776
          Stem-loop Sequence
          AGGGACGGGACGUGGUGCAGUGUUGUUCUUUCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028519
          miRBase ID: mdo-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-92b
          Stem-loop Accession # MI0034329
          Stem-loop Sequence
          CAGGUCCGGGACGUGGUGCGCUGUUGUCAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042497
          miRBase ID: nbr-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-92b
          Stem-loop Accession # MI0040200
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049451
          miRBase ID: nle-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-92b
          Stem-loop Accession # MI0034603
          Stem-loop Sequence
          AGGUCCGGGAUGUGGUGCACUGUUGUAAUAUCUCCCCUCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042694
          miRBase ID: oni-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-92b
          Stem-loop Accession # MI0039788
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049080
          miRBase ID: ppa-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-92b
          Stem-loop Accession # MI0020575
          Stem-loop Sequence
          AGCGGGAUCCCGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024028
          miRBase ID: ptr-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-92b
          Stem-loop Accession # MI0013126
          Stem-loop Sequence
          GCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013909
          miRBase ID: ssc-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-92b
          Stem-loop Accession # MI0014809
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUUUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCUCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015744
          miRBase ID: ppy-miR-92b
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-92b
          Stem-loop Accession # MI0032394
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUUCCCCGCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039908
          miRBase ID: pal-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-92b
          Stem-loop Accession # MI0007607
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006177
          miRBase ID: mml-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-92b
          Stem-loop Accession # MI0026762
          Stem-loop Sequence
          AGGUGUGGGAUGUGGUGCACUGUUGUACAAUGCCCUCCCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032712
          miRBase ID: ssa-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-92b
          Stem-loop Accession # MI0001953
          Stem-loop Sequence
          UCCUACGGGCAGGGAGGUGUGGGAUGUUGUGCAGUGUUGUUCAAUCUCCCGCCAAUAUUGCACUCGUCCCGGCCUCCCUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001809
          miRBase ID: dre-miR-92b-3p
          Mature miRNA Sequence: UAUUGCACUCGUCCCGGCCUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464732_pri
          TaqMan™ Pri-miRNA Assay : Hs03294072_pri
          TaqMan™ Pri-miRNA Assay : Mm03308629_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10102
          Ambion® Pre-miR™ miRNA Precursor : PM10102
          mirVana® miRNA inhibitor : MH10102
          mirVana® miRNA mimic : MC10102
          TaqMan™ Advanced miRNA Assay : rno481277_mir
          TaqMan™ Advanced miRNA Assay : mmu481277_mir
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 4
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 4
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards


          miRBase Alias:

          Danio rerio: dre-miR-92b (v20)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline