Search
Search

| Assay Name: | hsa-miR-369-5p |
|---|---|
| miRBase Accession Number: | MI0000777 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAUCGACCGUGUUAUAUUCGC |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAUCGACCGUGUUAUAUUCGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-369-5p | MIMAT0001621 |
| Mouse | mmu-miR-369-5p | MIMAT0003185 |
| Rat | rno-miR-369-5p | MIMAT0003206 |
| Cricetulus griseus | cgr-miR-369-5p | MIMAT0023937 |
| Pongo pygmaeus | ppy-miR-369-5p | MIMAT0015855 |
| Rhesus monkey | mml-miR-369-5p | MIMAT0006293 |