Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03466772_pri
          Assay Name:
          rno-mir-409b
          Stem-loop ID:
          rno-mir-3578 rno-mir-369 rno-mir-409a rno-mir-409b rno-mir-412
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.6: 133893672 - 133893788 [-] on Build Rnor_6.0
          Stem-loop Location:
          Chr.6: 133893672 - 133893788 [-] on Build Rnor_6.0 Chr.6: 133893693 - 133893771 [+] on Build Rnor_6.0 Chr.6: 133893419 - 133893495 [+] on Build Rnor_6.0 Chr.6: 133893418 - 133893497 [-] on Build Rnor_6.0 Chr.6: 133893571 - 133893640 [+] on Build Rnor_6.0
          Mature miRNA ID:
          rno-miR-3578 rno-miR-369-5p rno-miR-369-3p rno-miR-409a-5p rno-miR-409a-3p rno-miR-409b rno-miR-412-5p rno-miR-412-3p
          Species:
          Rat
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Rn03466772_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          62

          Chromosome Location:

          Chr. 6 - 133893278 - 133893278 [-] on 133893278 Build Rnor_6.0

          Species:

          Rat

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-3578 MIMAT0017866
          Rat rno-miR-369-5p MIMAT0003206
          Rat rno-miR-369-3p MIMAT0003207
          Rat rno-miR-409a-5p MIMAT0003204
          Rat rno-miR-409a-3p MIMAT0003205
          Rat rno-miR-409b MIMAT0017870
          View More Rows
          Rat rno-miR-412-5p MIMAT0017204
          Rat rno-miR-412-3p MIMAT0003124

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-3578
          Stem-loop Accession # MI0015451
          Stem-loop Sequence
          UCUCCAAGCUGAGCAUUGAUACUGAGAAAAGAUCAACCAUGUAUUAUUCGAAGUCAGCUAAGCGAAUAUAACACGGUCGAUCUCCCUUCAAGUACCAGCAGAGGUGCCGAGCUGAGG
          Chromosome Location Chr. 6 - 133893672 - 133893788 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017866
          miRBase ID: rno-miR-3578
          Mature miRNA Sequence: GCGAAUAUAACACGGUCGAUCU
          Chromosome Location: Chr. 6 - 133893672 - 133893788 [-] on Build Rnor_6.0

          Gene Family ID MIPF0001166, mir-3578
          Distance to Amplicon 363 bases
          Stem-loop ID rno-mir-369
          Stem-loop Accession # MI0003551
          Stem-loop Sequence
          GGUACUUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUAGCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUC
          Chromosome Location Chr. 6 - 133893693 - 133893771 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003206
          miRBase ID: rno-miR-369-5p
          Mature miRNA Sequence: AGAUCGACCGUGUUAUAUUCGC
          Chromosome Location: Chr. 6 - 133893693 - 133893771 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0003207
          miRBase ID: rno-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: Chr. 6 - 133893693 - 133893771 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000018, mir-154
          Distance to Amplicon 384 bases
          Stem-loop ID rno-mir-409a
          Stem-loop Accession # MI0003550
          Stem-loop Sequence
          GGUACUCGGAGAGAGGUUACCCGAGCAACUUUGCAUCUGGAGGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUC
          Chromosome Location Chr. 6 - 133893419 - 133893495 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003204
          miRBase ID: rno-miR-409a-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: Chr. 6 - 133893419 - 133893495 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0003205
          miRBase ID: rno-miR-409a-3p
          Mature miRNA Sequence: AAUGUUGCUCGGUGAACCCC
          Chromosome Location: Chr. 6 - 133893419 - 133893495 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000018, mir-154
          Distance to Amplicon 110 bases
          Stem-loop ID rno-mir-409b
          Stem-loop Accession # MI0015454
          Stem-loop Sequence
          UUGAUACCGAAAAGGGGUUCACCGAGCAACAUUCGUCCUCCAGAUGCAAAGUUGCUCGGGUAACCUCUCUCCGAGUACCA
          Chromosome Location Chr. 6 - 133893418 - 133893497 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017870
          miRBase ID: rno-miR-409b
          Mature miRNA Sequence: AGGGGUUCACCGAGCAACAUUCG
          Chromosome Location: Chr. 6 - 133893418 - 133893497 [-] on Build Rnor_6.0

          Gene Family ID MIPF0000018, mir-154
          Distance to Amplicon 109 bases
          Stem-loop ID rno-mir-412
          Stem-loop Accession # MI0003488
          Stem-loop Sequence
          GGGACGGAUGGUCGACCAGCUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGCCGUCGGU
          Chromosome Location Chr. 6 - 133893571 - 133893640 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017204
          miRBase ID: rno-miR-412-5p
          Mature miRNA Sequence: UGGUCGACCAGCUGGAAAGU
          Chromosome Location: Chr. 6 - 133893571 - 133893640 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0003124
          miRBase ID: rno-miR-412-3p
          Mature miRNA Sequence: CUUCACCUGGUCCACUAGCCGU
          Chromosome Location: Chr. 6 - 133893571 - 133893640 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000192, mir-412
          Distance to Amplicon 262 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM20536
          Ambion® Pre-miR™ miRNA Precursor : PM20536
          TaqMan™ MicroRNA Assay : 465096_mat
          mirVana® miRNA inhibitor : MH20536
          mirVana® miRNA mimic : MC20536
          TaqMan™ Advanced miRNA Assay : rno481392_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19879
          Ambion® Pre-miR™ miRNA Precursor : PM19879
          TaqMan™ MicroRNA Assay : 463026_mat
          mirVana® miRNA inhibitor : MH19879
          mirVana® miRNA mimic : MC19879
          TaqMan™ Advanced miRNA Assay : rno481158_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20054
          Ambion® Pre-miR™ miRNA Precursor : PM20054
          TaqMan™ MicroRNA Assay : 464677_mat
          mirVana® miRNA inhibitor : MH20054
          mirVana® miRNA mimic : MC20054
          TaqMan™ Advanced miRNA Assay : rno481114_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13028
          Ambion® Pre-miR™ miRNA Precursor : PM13028
          TaqMan™ MicroRNA Assay : 002331
          mirVana® miRNA inhibitor : MH13028
          mirVana® miRNA mimic : MC13028
          TaqMan™ Advanced miRNA Assay : rno478872_mir
          TaqMan™ Advanced miRNA Assay : mmu478872_mir
          TaqMan™ Advanced miRNA Assay : 478872_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11303
          Ambion® Pre-miR™ miRNA Precursor : PM11303
          TaqMan™ MicroRNA Assay : 002679
          mirVana® miRNA inhibitor : MH11303
          mirVana® miRNA mimic : MC11303
          TaqMan™ Advanced miRNA Assay : rno481154_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10963
          Ambion® Pre-miR™ miRNA Precursor : PM10963
          TaqMan™ MicroRNA Assay : 001021
          mirVana® miRNA inhibitor : MH10963
          mirVana® miRNA mimic : MC10963
          TaqMan™ Advanced miRNA Assay : mmu481129_mir
          TaqMan™ Advanced miRNA Assay : rno481129_mir
          TaqMan™ Advanced miRNA Assay : 478068_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10825
          Ambion® Pre-miR™ miRNA Precursor : PM10825
          TaqMan™ MicroRNA Assay : 000557
          mirVana® miRNA inhibitor : MH10825
          mirVana® miRNA mimic : MC10825
          TaqMan™ Advanced miRNA Assay : mmu481386_mir
          TaqMan™ Advanced miRNA Assay : 478067_mir
          TaqMan™ Advanced miRNA Assay : rno481386_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20480
          Ambion® Pre-miR™ miRNA Precursor : PM20480
          TaqMan™ MicroRNA Assay : 463694_mat
          mirVana® miRNA inhibitor : MH20480
          mirVana® miRNA mimic : MC20480
          TaqMan™ Advanced miRNA Assay : rno481155_mir


          miRBase Alias:

          Rat: rno-mir-369 (v21)
          Rat: rno-mir-409a (v21)
          Rat: rno-mir-412 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline