Search
Search

| Assay Name: | hsa-miR-409-5p |
|---|---|
| Stem-loop Accession Number: | MI0001735 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUUACCCGAGCAACUUUGCAU |
| Species: | Human, Mouse, Rat, Bovine, Goat, Horse, Nomascus leucogenys, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sheep |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUACCCGAGCAACUUUGCAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-409-5p | MIMAT0001638 |
| Mouse | mmu-miR-409-5p | MIMAT0004746 |
| Rat | rno-miR-409a-5p | MIMAT0003204 |
| Bovine | bta-miR-409a | MIMAT0009310 |
| Goat | chi-miR-409-5p | MIMAT0036209 |
| Horse | eca-miR-409-5p | MIMAT0013151 |
| View More Rows | ||
| Nomascus leucogenys | nle-miR-409 | MIMAT0049370 |
| Pongo pygmaeus | ppy-miR-409-5p | MIMAT0015875 |
| Pteropus alecto | pal-miR-409-5p | MIMAT0040126 |
| Rhesus monkey | mml-miR-409-5p | MIMAT0006314 |
| Sheep | oar-miR-409-5p | MIMAT0019327 |