Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13028
          Assay Name: hsa-miR-409-5p
          miRBase Accession Number: MI0001735
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065300 - 101065378 [+] on Build GRCh38
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Species: Human, Mouse, Rat, Bovine, Goat, Horse, Nomascus leucogenys, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA mimic
          Assay ID MC13028

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AGGUUACCCGAGCAACUUUGCAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-409-5p MIMAT0001638
          Mouse mmu-miR-409-5p MIMAT0004746
          Rat rno-miR-409a-5p MIMAT0003204
          Bovine bta-miR-409a MIMAT0009310
          Goat chi-miR-409-5p MIMAT0036209
          Horse eca-miR-409-5p MIMAT0013151
          View More Rows
          Nomascus leucogenys nle-miR-409 MIMAT0049370
          Pongo pygmaeus ppy-miR-409-5p MIMAT0015875
          Pteropus alecto pal-miR-409-5p MIMAT0040126
          Rhesus monkey mml-miR-409-5p MIMAT0006314
          Sheep oar-miR-409-5p MIMAT0019327

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-409
          Stem-loop Accession # MI0001735
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location Chr. 14 - 101065300 - 101065378 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001638
          miRBase ID: hsa-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: Chr. 14 - 101065300 - 101065378 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-409
          Stem-loop Accession # MI0001160
          Stem-loop Sequence
          UGGUACUCGGAGAGAGGUUACCCGAGCAACUUUGCAUCUGGAGGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004746
          miRBase ID: mmu-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-409a
          Stem-loop Accession # MI0003550
          Stem-loop Sequence
          GGUACUCGGAGAGAGGUUACCCGAGCAACUUUGCAUCUGGAGGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUC
          Chromosome Location Chr. 6 - 133893419 - 133893495 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003204
          miRBase ID: rno-miR-409a-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: Chr. 6 - 133893419 - 133893495 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-409a
          Stem-loop Accession # MI0009824
          Stem-loop Sequence
          UGGUACGCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009310
          miRBase ID: bta-miR-409a
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-409
          Stem-loop Accession # MI0030782
          Stem-loop Sequence
          CCUCUCCAUGGUACGCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCAAAAUCCAUCAGAGAGGCCAUCUGGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036209
          miRBase ID: chi-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-409
          Stem-loop Accession # MI0012896
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013151
          miRBase ID: eca-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-409
          Stem-loop Accession # MI0040107
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049370
          miRBase ID: nle-miR-409
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-409
          Stem-loop Accession # MI0014933
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015875
          miRBase ID: ppy-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-409
          Stem-loop Accession # MI0032507
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040126
          miRBase ID: pal-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-409
          Stem-loop Accession # MI0007733
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006314
          miRBase ID: mml-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-409
          Stem-loop Accession # MI0016958
          Stem-loop Sequence
          CCUCUCCAUGGUACGCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCAAAAUCCAUCAGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019327
          miRBase ID: oar-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466772_pri
          TaqMan™ Pri-miRNA Assay : Mm03307706_pri
          TaqMan™ Pri-miRNA Assay : Hs04225933_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13028
          Ambion® Pre-miR™ miRNA Precursor : PM13028
          TaqMan™ MicroRNA Assay : 002331
          mirVana® miRNA inhibitor : MH13028
          TaqMan™ Advanced miRNA Assay : rno478872_mir
          TaqMan™ Advanced miRNA Assay : mmu478872_mir
          TaqMan™ Advanced miRNA Assay : 478872_mir


          miRBase Alias:

          Rat: rno-miR-409-5p (v18)
          Bos taurus: bta-miR-409 (v16)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline