Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03307706_pri
          Assay Name:
          mmu-mir-409
          Stem-loop ID:
          mmu-mir-369 mmu-mir-409 mmu-mir-412
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.12: 109743418 - 109743496 [+] on Build GRCm38
          Stem-loop Location:
          Chr.12: 109743418 - 109743496 [+] on Build GRCm38 Chr.12: 109743158 - 109743236 [+] on Build GRCm38 Chr.12: 109743289 - 109743368 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-369-5p mmu-miR-369-3p mmu-miR-409-5p mmu-miR-409-3p mmu-miR-412-5p mmu-miR-412-3p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03307706_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          61

          Chromosome Location:

          Chr. 12 - 109743017 - 109743017 [-] on 109743017 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-369-5p MIMAT0003185
          Mouse mmu-miR-369-3p MIMAT0003186
          Mouse mmu-miR-409-5p MIMAT0004746
          Mouse mmu-miR-409-3p MIMAT0001090
          Mouse mmu-miR-412-5p MIMAT0017173
          Mouse mmu-miR-412-3p MIMAT0001094

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-369
          Stem-loop Accession # MI0003535
          Stem-loop Sequence
          GGUACUUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUGGCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUC
          Chromosome Location Chr. 12 - 109743418 - 109743496 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003185
          miRBase ID: mmu-miR-369-5p
          Mature miRNA Sequence: AGAUCGACCGUGUUAUAUUCGC
          Chromosome Location: Chr. 12 - 109743418 - 109743496 [+] on Build GRCm38

          miRBase Accession #: MIMAT0003186
          miRBase ID: mmu-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: Chr. 12 - 109743418 - 109743496 [+] on Build GRCm38

          Gene Family ID MIPF0000018, mir-154
          Distance to Amplicon 371 bases
          Stem-loop ID mmu-mir-409
          Stem-loop Accession # MI0001160
          Stem-loop Sequence
          UGGUACUCGGAGAGAGGUUACCCGAGCAACUUUGCAUCUGGAGGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004746
          miRBase ID: mmu-miR-409-5p
          Mature miRNA Sequence: AGGUUACCCGAGCAACUUUGCAU
          Chromosome Location: Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38

          miRBase Accession #: MIMAT0001090
          miRBase ID: mmu-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38

          Gene Family ID MIPF0000018, mir-154
          Distance to Amplicon 111 bases
          Stem-loop ID mmu-mir-412
          Stem-loop Accession # MI0001164
          Stem-loop Sequence
          GGGUAUGGGACGGAUGGUCGACCAGCUGGAAAGUAAUUGUUUCUAAUGUACUUCACCUGGUCCACUAGCCGUCGGUGCCC
          Chromosome Location Chr. 12 - 109743289 - 109743368 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017173
          miRBase ID: mmu-miR-412-5p
          Mature miRNA Sequence: UGGUCGACCAGCUGGAAAGUAAU
          Chromosome Location: Chr. 12 - 109743289 - 109743368 [+] on Build GRCm38

          miRBase Accession #: MIMAT0001094
          miRBase ID: mmu-miR-412-3p
          Mature miRNA Sequence: UUCACCUGGUCCACUAGCCG
          Chromosome Location: Chr. 12 - 109743289 - 109743368 [+] on Build GRCm38

          Gene Family ID MIPF0000192, mir-412
          Distance to Amplicon 242 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM19597
          Ambion® Pre-miR™ miRNA Precursor : PM19597
          TaqMan™ MicroRNA Assay : 464537_mat
          mirVana® miRNA inhibitor : MH19597
          mirVana® miRNA mimic : MC19597
          TaqMan™ Advanced miRNA Assay : mmu482761_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12722
          Ambion® Pre-miR™ miRNA Precursor : PM12722
          TaqMan™ MicroRNA Assay : 002575
          mirVana® miRNA inhibitor : MH12722
          mirVana® miRNA mimic : MC12722
          TaqMan™ Advanced miRNA Assay : mmu481832_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10963
          Ambion® Pre-miR™ miRNA Precursor : PM10963
          TaqMan™ MicroRNA Assay : 001021
          mirVana® miRNA inhibitor : MH10963
          mirVana® miRNA mimic : MC10963
          TaqMan™ Advanced miRNA Assay : mmu481129_mir
          TaqMan™ Advanced miRNA Assay : rno481129_mir
          TaqMan™ Advanced miRNA Assay : 478068_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10825
          Ambion® Pre-miR™ miRNA Precursor : PM10825
          TaqMan™ MicroRNA Assay : 000557
          mirVana® miRNA inhibitor : MH10825
          mirVana® miRNA mimic : MC10825
          TaqMan™ Advanced miRNA Assay : mmu481386_mir
          TaqMan™ Advanced miRNA Assay : 478067_mir
          TaqMan™ Advanced miRNA Assay : rno481386_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13028
          Ambion® Pre-miR™ miRNA Precursor : PM13028
          TaqMan™ MicroRNA Assay : 002331
          mirVana® miRNA inhibitor : MH13028
          mirVana® miRNA mimic : MC13028
          TaqMan™ Advanced miRNA Assay : rno478872_mir
          TaqMan™ Advanced miRNA Assay : mmu478872_mir
          TaqMan™ Advanced miRNA Assay : 478872_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12446
          Ambion® Pre-miR™ miRNA Precursor : PM12446
          TaqMan™ MicroRNA Assay : 002332
          mirVana® miRNA inhibitor : MH12446
          mirVana® miRNA mimic : MC12446
          TaqMan™ Advanced miRNA Assay : mmu481831_mir
          TaqMan™ Advanced miRNA Assay : 478084_mir


          miRBase Alias:

          Mouse: mmu-mir-369 (v21)
          Mouse: mmu-mir-412 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline