Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12446
          Assay Name: hsa-miR-409-3p
          miRBase Accession Number: MI0001735
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065300 - 101065378 [+] on Build GRCh38
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Species: Human, Mouse, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12446

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          GAAUGUUGCUCGGUGAACCCCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-409-3p MIMAT0001639
          Mouse mmu-miR-409-3p MIMAT0001090
          Callithrix jacchus cja-miR-409 MIMAT0049324
          Cricetulus griseus cgr-miR-409-3p MIMAT0023951
          Dasypus novemcinctus dno-miR-409-3p MIMAT0047871
          Guinea pig cpo-miR-409-3p MIMAT0047234
          View More Rows
          Horse eca-miR-409-3p MIMAT0013152
          Pan paniscus ppa-miR-409 MIMAT0049036
          Pan troglodytes ptr-miR-409 MIMAT0008136
          Papio hamadryas pha-miR-409 MIMAT0049576
          Pongo pygmaeus ppy-miR-409-3p MIMAT0015876
          Pteropus alecto pal-miR-409-3p MIMAT0040127
          Rabbit ocu-miR-409-3p MIMAT0048434
          Rhesus monkey mml-miR-409-3p MIMAT0006315
          Tupaia chinensis tch-miR-409-3p MIMAT0036502

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-409
          Stem-loop Accession # MI0001735
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location Chr. 14 - 101065300 - 101065378 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001639
          miRBase ID: hsa-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: Chr. 14 - 101065300 - 101065378 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-409
          Stem-loop Accession # MI0001160
          Stem-loop Sequence
          UGGUACUCGGAGAGAGGUUACCCGAGCAACUUUGCAUCUGGAGGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001090
          miRBase ID: mmu-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: Chr. 12 - 109743158 - 109743236 [+] on Build GRCm38

          Callithrix jacchus

          Stem-loop ID cja-mir-409
          Stem-loop Accession # MI0040060
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUAGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049324
          miRBase ID: cja-miR-409
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-409
          Stem-loop Accession # MI0020520
          Stem-loop Sequence
          CUCGGAGAGAGGUUACCCGAGCAACUUUGCAUCUGGAGGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023951
          miRBase ID: cgr-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-409
          Stem-loop Accession # MI0039100
          Stem-loop Sequence
          UGGUACGCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUCUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047871
          miRBase ID: dno-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-409
          Stem-loop Accession # MI0038765
          Stem-loop Sequence
          AGGUUACCCAAGCAACUUUGCAUCUGAACAACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047234
          miRBase ID: cpo-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-409
          Stem-loop Accession # MI0012896
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013152
          miRBase ID: eca-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-409
          Stem-loop Accession # MI0039744
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049036
          miRBase ID: ppa-miR-409
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-409
          Stem-loop Accession # MI0008657
          Stem-loop Sequence
          GGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008136
          miRBase ID: ptr-miR-409
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-409
          Stem-loop Accession # MI0040351
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049576
          miRBase ID: pha-miR-409
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-409
          Stem-loop Accession # MI0014933
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015876
          miRBase ID: ppy-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-409
          Stem-loop Accession # MI0032507
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040127
          miRBase ID: pal-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-409
          Stem-loop Accession # MI0039398
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048434
          miRBase ID: ocu-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-409
          Stem-loop Accession # MI0007733
          Stem-loop Sequence
          UGGUACUCGGGGAGAGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCUUUUCGGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006315
          miRBase ID: mml-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-409
          Stem-loop Accession # MI0031159
          Stem-loop Sequence
          AGGUUACCCGAGCAACUUUGCAUCUGGACGACGAAUGUUGCUCGGUGAACCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036502
          miRBase ID: tch-miR-409-3p
          Mature miRNA Sequence: GAAUGUUGCUCGGUGAACCCCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307706_pri
          TaqMan™ Pri-miRNA Assay : Hs04225933_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12446
          Ambion® Pre-miR™ miRNA Precursor : PM12446
          TaqMan™ MicroRNA Assay : 002332
          mirVana® miRNA mimic : MC12446
          TaqMan™ Advanced miRNA Assay : mmu481831_mir
          TaqMan™ Advanced miRNA Assay : 478084_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline