Search
Search

| Assay Name: | hsa-miR-409-3p |
|---|---|
| miRBase Accession Number: | MI0001735 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAAUGUUGCUCGGUGAACCCCU |
| Species: | Human, Mouse, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GAAUGUUGCUCGGUGAACCCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-409-3p | MIMAT0001639 |
| Mouse | mmu-miR-409-3p | MIMAT0001090 |
| Callithrix jacchus | cja-miR-409 | MIMAT0049324 |
| Cricetulus griseus | cgr-miR-409-3p | MIMAT0023951 |
| Dasypus novemcinctus | dno-miR-409-3p | MIMAT0047871 |
| Guinea pig | cpo-miR-409-3p | MIMAT0047234 |
| View More Rows | ||
| Horse | eca-miR-409-3p | MIMAT0013152 |
| Pan paniscus | ppa-miR-409 | MIMAT0049036 |
| Pan troglodytes | ptr-miR-409 | MIMAT0008136 |
| Papio hamadryas | pha-miR-409 | MIMAT0049576 |
| Pongo pygmaeus | ppy-miR-409-3p | MIMAT0015876 |
| Pteropus alecto | pal-miR-409-3p | MIMAT0040127 |
| Rabbit | ocu-miR-409-3p | MIMAT0048434 |
| Rhesus monkey | mml-miR-409-3p | MIMAT0006315 |
| Tupaia chinensis | tch-miR-409-3p | MIMAT0036502 |