Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481386_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-369-3p
          Stem-loop Accession Number: MI0000777
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101065598 - 101065667 [+] on Build GRCh38
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Goat, Gorilla gorilla, Guinea pig, Horse, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sheep, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481386_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AAUAAUACAUGGUUGAUCUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-369-3p MIMAT0000721
          Mouse mmu-miR-369-3p MIMAT0003186
          Rat rno-miR-369-3p MIMAT0003207
          Bovine bta-miR-369-3p MIMAT0003802
          Callithrix jacchus cja-miR-369-3p MIMAT0039353
          Cricetulus griseus cgr-miR-369-3p MIMAT0023938
          View More Rows
          Dasypus novemcinctus dno-miR-369-3p MIMAT0047841
          Goat chi-miR-369-3p MIMAT0036179
          Gorilla gorilla ggo-miR-369 MIMAT0049114
          Guinea pig cpo-miR-369-3p MIMAT0047200
          Horse eca-miR-369-3p MIMAT0013141
          Pan paniscus ppa-miR-369 MIMAT0049034
          Pan troglodytes ptr-miR-369 MIMAT0008119
          Pig ssc-miR-369 MIMAT0015714
          Pongo pygmaeus ppy-miR-369-3p MIMAT0015856
          Pteropus alecto pal-miR-369-3p MIMAT0040024
          Rhesus monkey mml-miR-369-3p MIMAT0006294
          Sheep oar-miR-369-3p MIMAT0019332
          Tupaia chinensis tch-miR-369-3p MIMAT0036544

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-369
          Stem-loop Accession # MI0000777
          Stem-loop Sequence
          UUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAG
          Chromosome Location Chr. 14 - 101065598 - 101065667 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000721
          miRBase ID: hsa-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: Chr. 14 - 101065598 - 101065667 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-369
          Stem-loop Accession # MI0003535
          Stem-loop Sequence
          GGUACUUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUGGCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUC
          Chromosome Location Chr. 12 - 109743418 - 109743496 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003186
          miRBase ID: mmu-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: Chr. 12 - 109743418 - 109743496 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-369
          Stem-loop Accession # MI0003551
          Stem-loop Sequence
          GGUACUUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUAGCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUC
          Chromosome Location Chr. 6 - 133893693 - 133893771 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003207
          miRBase ID: rno-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: Chr. 6 - 133893693 - 133893771 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-369
          Stem-loop Accession # MI0005020
          Stem-loop Sequence
          UGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003802
          miRBase ID: bta-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-369
          Stem-loop Accession # MI0031867
          Stem-loop Sequence
          UGAAGAGAGAUCGACCGUGUUAUAUUCGCUUUGUUGAUUUCGAAUAAUACAUGGUUGAUCUUUUCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039353
          miRBase ID: cja-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-369
          Stem-loop Accession # MI0020512
          Stem-loop Sequence
          CUUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUGGCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCGGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023938
          miRBase ID: cgr-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-369
          Stem-loop Accession # MI0039085
          Stem-loop Sequence
          AGAUCGACCGUGUUAUAUUCGCUUUCCUGACUUCGAAUAAUACAUGGUUGAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047841
          miRBase ID: dno-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-369
          Stem-loop Accession # MI0030764
          Stem-loop Sequence
          AGCAUCUCUCCUGGUACCUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUCAAAUCUCACCUUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036179
          miRBase ID: chi-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-369
          Stem-loop Accession # MI0039826
          Stem-loop Sequence
          AGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049114
          miRBase ID: ggo-miR-369
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-369
          Stem-loop Accession # MI0038748
          Stem-loop Sequence
          AGAUCGACCGUGUUAUAUUCGCUUCCUUGACUUCGAAUAAUACAUGGUUGAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047200
          miRBase ID: cpo-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-369
          Stem-loop Accession # MI0012886
          Stem-loop Sequence
          UUGAAGGGAGAUCGACCGUGUCAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013141
          miRBase ID: eca-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-369
          Stem-loop Accession # MI0039742
          Stem-loop Sequence
          AGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049034
          miRBase ID: ppa-miR-369
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-369
          Stem-loop Accession # MI0008639
          Stem-loop Sequence
          UGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008119
          miRBase ID: ptr-miR-369
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-369
          Stem-loop Accession # MI0014774
          Stem-loop Sequence
          UUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUGGUACUUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUGGCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUCUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015714
          miRBase ID: ssc-miR-369
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-369
          Stem-loop Accession # MI0014914
          Stem-loop Sequence
          UUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015856
          miRBase ID: ppy-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-369
          Stem-loop Accession # MI0032453
          Stem-loop Sequence
          AGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040024
          miRBase ID: pal-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-369
          Stem-loop Accession # MI0007713
          Stem-loop Sequence
          UUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006294
          miRBase ID: mml-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-369
          Stem-loop Accession # MI0016960
          Stem-loop Sequence
          AGCAUCUCUCCUGGUACCUGAAGGGAGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUUUCUCAGUAUCAAAUCUCACCUUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019332
          miRBase ID: oar-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-369
          Stem-loop Accession # MI0031204
          Stem-loop Sequence
          AGAUCGACCGUGUUAUAUUCGCUUUAUUGACUUCGAAUAAUACAUGGUUGAUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036544
          miRBase ID: tch-miR-369-3p
          Mature miRNA Sequence: AAUAAUACAUGGUUGAUCUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307706_pri
          TaqMan™ Pri-miRNA Assay : Rn03466772_pri
          TaqMan™ Pri-miRNA Assay : Hs04225933_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10825
          Ambion® Pre-miR™ miRNA Precursor : PM10825
          TaqMan™ MicroRNA Assay : 000557
          mirVana® miRNA inhibitor : MH10825
          mirVana® miRNA mimic : MC10825
          TaqMan™ Advanced miRNA Assay : 478067_mir
          TaqMan™ Advanced miRNA Assay : rno481386_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline