Search
Search

| Assay Name: | hsa-miR-369-3p |
|---|---|
| miRBase Accession Number: | MI0000777 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAUAAUACAUGGUUGAUCUUU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Goat, Gorilla gorilla, Guinea pig, Horse, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sheep, Tupaia chinensis |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUAAUACAUGGUUGAUCUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-369-3p | MIMAT0000721 |
| Mouse | mmu-miR-369-3p | MIMAT0003186 |
| Rat | rno-miR-369-3p | MIMAT0003207 |
| Bovine | bta-miR-369-3p | MIMAT0003802 |
| Callithrix jacchus | cja-miR-369-3p | MIMAT0039353 |
| Cricetulus griseus | cgr-miR-369-3p | MIMAT0023938 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-369-3p | MIMAT0047841 |
| Goat | chi-miR-369-3p | MIMAT0036179 |
| Gorilla gorilla | ggo-miR-369 | MIMAT0049114 |
| Guinea pig | cpo-miR-369-3p | MIMAT0047200 |
| Horse | eca-miR-369-3p | MIMAT0013141 |
| Pan paniscus | ppa-miR-369 | MIMAT0049034 |
| Pan troglodytes | ptr-miR-369 | MIMAT0008119 |
| Pig | ssc-miR-369 | MIMAT0015714 |
| Pongo pygmaeus | ppy-miR-369-3p | MIMAT0015856 |
| Pteropus alecto | pal-miR-369-3p | MIMAT0040024 |
| Rhesus monkey | mml-miR-369-3p | MIMAT0006294 |
| Sheep | oar-miR-369-3p | MIMAT0019332 |
| Tupaia chinensis | tch-miR-369-3p | MIMAT0036544 |