Search
Search

| Assay Name: | mmu-miR-412-3p |
|---|---|
| miRBase Accession Number: | MI0001164 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCACCUGGUCCACUAGCCG |
| Species: | Mouse, Horse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000192, mir-412 |
Mature miRNA Sequence: | UUCACCUGGUCCACUAGCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-412-3p | MIMAT0001094 |
| Horse | eca-miR-412 | MIMAT0013155 |