Search
Search

| Assay Name: | rno-miR-3578 |
|---|---|
| miRBase Accession Number: | MI0015451 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCGAAUAUAACACGGUCGAUCU |
| Species: | Rat |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001166, mir-3578 |
Mature miRNA Sequence: | GCGAAUAUAACACGGUCGAUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3578 | MIMAT0017866 |