Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03303133_pri
          Assay Name:
          hsa-mir-135a-2
          Stem-loop ID:
          hsa-mir-135a-2
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.12: 97563812 - 97563911 [+] on Build GRCh38
          Stem-loop Location:
          Chr.12: 97563812 - 97563911 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-135a-5p hsa-miR-135a-2-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03303133_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          76

          Chromosome Location:

          Chr. 12 - 97563513 - 97563513 [+] on 97563513 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-135a-5p MIMAT0000428
          Human hsa-miR-135a-2-3p MIMAT0037309

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-135a-2
          Stem-loop Accession # MI0000453
          Stem-loop Sequence
          AGAUAAAUUCACUCUAGUGCUUUAUGGCUUUUUAUUCCUAUGUGAUAGUAAUAAAGUCUCAUGUAGGGAUGGAAGCCAUGAAAUACAUUGUGAAAAAUCA
          Chromosome Location Chr. 12 - 97563812 - 97563911 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000428
          miRBase ID: hsa-miR-135a-5p
          Mature miRNA Sequence: UAUGGCUUUUUAUUCCUAUGUGA
          Chromosome Location: Chr. 12 - 97563812 - 97563911 [+] on Build GRCh38

          miRBase Accession #: MIMAT0037309
          miRBase ID: hsa-miR-135a-2-3p
          Mature miRNA Sequence: AUGUAGGGAUGGAAGCCAUGAA
          Chromosome Location: Chr. 12 - 97563812 - 97563911 [+] on Build GRCh38

          Gene Family ID MIPF0000028, mir-135
          Distance to Amplicon 262 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11126
          Ambion® Pre-miR™ miRNA Precursor : PM11126
          TaqMan™ MicroRNA Assay : 000460
          mirVana® miRNA inhibitor : MH11126
          mirVana® miRNA mimic : MC11126
          TaqMan™ Advanced miRNA Assay : 478581_mir
          TaqMan™ Advanced miRNA Assay : rno478581_mir
          TaqMan™ Advanced miRNA Assay : mmu478581_mir
          TaqMan™ MicroRNA Assay : 477768_mat
          mirVana® miRNA inhibitor : MH23887
          mirVana® miRNA mimic : MC23887
          TaqMan™ Advanced miRNA Assay : 483071_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline