Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04257850_pri
          Assay Name:
          hsa-mir-4761
          Stem-loop ID:
          hsa-mir-4761
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.22: 19963753 - 19963834 [+] on Build GRCh38
          Stem-loop Location:
          Chr.22: 19963753 - 19963834 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-4761-5p hsa-miR-4761-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs04257850_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          119

          Chromosome Location:

          Chr. 22 - 19963604 - 19963604 [-] on 19963604 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-4761-5p MIMAT0019908
          Human hsa-miR-4761-3p MIMAT0019909

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-4761
          Stem-loop Accession # MI0017402
          Stem-loop Sequence
          GGACAAGGUGUGCAUGCCUGACCCGUUGUCAGACCUGGAAAAAGGGCCGGCUGUGGGCAGGGAGGGCAUGCGCACUUUGUCC
          Chromosome Location Chr. 22 - 19963753 - 19963834 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0019908
          miRBase ID: hsa-miR-4761-5p
          Mature miRNA Sequence: ACAAGGUGUGCAUGCCUGACC
          Chromosome Location: Chr. 22 - 19963753 - 19963834 [+] on Build GRCh38

          miRBase Accession #: MIMAT0019909
          miRBase ID: hsa-miR-4761-3p
          Mature miRNA Sequence: GAGGGCAUGCGCACUUUGUCC
          Chromosome Location: Chr. 22 - 19963753 - 19963834 [+] on Build GRCh38

          Gene Family ID
          Distance to Amplicon 106 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM22718
          Ambion® Pre-miR™ miRNA Precursor : PM22718
          TaqMan™ MicroRNA Assay : 463092_mat
          mirVana® miRNA inhibitor : MH22718
          mirVana® miRNA mimic : MC22718
          TaqMan™ Advanced miRNA Assay : 480020_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM21740
          Ambion® Pre-miR™ miRNA Precursor : PM21740
          TaqMan™ MicroRNA Assay : 463379_mat
          mirVana® miRNA inhibitor : MH21740
          mirVana® miRNA mimic : MC21740
          TaqMan™ Advanced miRNA Assay : 480019_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline