Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn03465230_pri
Assay Name:
rno-mir-26a
Stem-loop ID:
rno-mir-26a
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.8: 127714441 - 127714530 [+] on Build Rnor_6.0
Stem-loop Location:
Chr.8: 127714441 - 127714530 [+] on Build Rnor_6.0
Mature miRNA ID:
rno-miR-26a-5p
rno-miR-26a-3p
Species:
Rat
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Rn03465230_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
55
Chromosome Location:
Chr. 8 - 127714196 - 127714196 [-] on 127714196 Build Rnor_6.0
Species:
Rat
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Rat
rno-miR-26a-5p
MIMAT0000796
Rat
rno-miR-26a-3p
MIMAT0017100
Stem-loop Details
Rat
Stem-loop ID
rno-mir-26a
Stem-loop Accession #
MI0000857
Stem-loop Sequence
AAGGCCGUGGCCUUGUUCAAGUAAUCCAGGAUAGGCUGUGCAGGUCCCAAGGGGCCUAUUCUUGGUUACUUGCACGGGGACGCGGGCCUG
Chromosome Location
Chr. 8 - 127714441 - 127714530 [+] on Build Rnor_6.0
Mature MicroRNA
miRBase Accession #: MIMAT0000796
miRBase ID: rno-miR-26a-5p
Mature miRNA Sequence: UUCAAGUAAUCCAGGAUAGGCU
Chromosome Location: Chr. 8 - 127714441 - 127714530 [+] on Build Rnor_6.0
miRBase Accession #: MIMAT0017100
miRBase ID: rno-miR-26a-3p
Mature miRNA Sequence: CCUAUUCUUGGUUACUUGCAC
Chromosome Location: Chr. 8 - 127714441 - 127714530 [+] on Build Rnor_6.0
Gene Family ID
MIPF0000043,
mir-26
Distance to Amplicon
218 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10249
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10249
TaqMan
™
MicroRNA Assay :
000405
mir
Vana
®
miRNA inhibitor :
MH10249
mir
Vana
®
miRNA mimic :
MC10249
TaqMan
™
Advanced miRNA Assay :
mmu481013_mir
TaqMan
™
Advanced miRNA Assay :
477995_mir
TaqMan
™
Advanced miRNA Assay :
rno481013_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM20611
Ambion
®
Pre-miR
™
miRNA Precursor :
PM20611
TaqMan
™
MicroRNA Assay :
462650_mat
mir
Vana
®
miRNA inhibitor :
MH20611
mir
Vana
®
miRNA mimic :
MC20611
TaqMan
™
Advanced miRNA Assay :
rno481012_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline