Search
Search

| Assay Name: | mmu-miR-1896 |
|---|---|
| Stem-loop Accession Number: | MI0008322 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCUCUGAUGGUGGGUGAGGAG |
| Species: | Mouse |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001981, mir-1896 |
Mature miRNA Sequence: | CUCUCUGAUGGUGGGUGAGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1896 | MIMAT0007873 |