Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480901_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-124-3p
          Stem-loop Accession Number: MI0000716
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 64590657 - 64590741 [+] on Build GRCm38
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Species: Mouse, Rat, Astatotilapia burtoni, Caenorhabditis remanei, Goat, Horse, Ictalurus punctatus, Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Polistes canadensis, Pongo pygmaeus, Pristionchus pacificus
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480901_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000021, mir-124

          Mature miRNA Sequence:

          UAAGGCACGCGGUGAAUGCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-124-3p MIMAT0000134
          Rat rno-miR-124-3p MIMAT0000828
          Astatotilapia burtoni abu-miR-124 MIMAT0042170
          Caenorhabditis remanei crm-miR-124a MIMAT0011541
          Goat chi-miR-124a MIMAT0035922
          Horse eca-miR-124 MIMAT0012906
          View More Rows
          Ictalurus punctatus ipu-miR-124a MIMAT0029396
          Metriaclima zebra mze-miR-124 MIMAT0042399
          Neolamprologus brichardi nbr-miR-124 MIMAT0042577
          Oreochromis niloticus oni-miR-124a MIMAT0042770
          Polistes canadensis pca-miR-124-3p MIMAT0045864
          Pongo pygmaeus ppy-miR-124 MIMAT0015747
          Pristionchus pacificus ppc-miR-124 MIMAT0011645

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-124-1
          Stem-loop Accession # MI0000716
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000134
          miRBase ID: mmu-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Stem-loop ID mmu-mir-124-2
          Stem-loop Accession # MI0000717
          Stem-loop Sequence
          AUCAAGAUCAGAGACUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 3 - 17795662 - 17795770 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000134
          miRBase ID: mmu-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Stem-loop ID mmu-mir-124-3
          Stem-loop Accession # MI0000150
          Stem-loop Sequence
          CUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAG
          Chromosome Location Chr. 2 - 180894040 - 180894107 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000134
          miRBase ID: mmu-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-124-1
          Stem-loop Accession # MI0000893
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 15 - 47964298 - 47964382 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000828
          miRBase ID: rno-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-124-2
          Stem-loop Accession # MI0000894
          Stem-loop Sequence
          AUCAAGAUCAGAGACUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000828
          miRBase ID: rno-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-124-3
          Stem-loop Accession # MI0000892
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location Chr. 3 - 176406205 - 176406291 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000828
          miRBase ID: rno-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-124-1
          Stem-loop Accession # MI0033894
          Stem-loop Sequence
          CGUGUUCACGGCGGACCUUGAUUUAUUAGCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042170
          miRBase ID: abu-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID abu-mir-124-2
          Stem-loop Accession # MI0033896
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042170
          miRBase ID: abu-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID abu-mir-124-3
          Stem-loop Accession # MI0033909
          Stem-loop Sequence
          UGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042170
          miRBase ID: abu-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID abu-mir-124-4
          Stem-loop Accession # MI0033910
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042170
          miRBase ID: abu-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID abu-mir-124-5
          Stem-loop Accession # MI0033912
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042170
          miRBase ID: abu-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID abu-mir-124-6
          Stem-loop Accession # MI0033985
          Stem-loop Sequence
          AGUGUUCACAGUGGACCUUGAUUUAAAAUCAGUCAAUUAAGGCACGCGGUGAAUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042170
          miRBase ID: abu-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Caenorhabditis remanei

          Stem-loop ID crm-mir-124a
          Stem-loop Accession # MI0011060
          Stem-loop Sequence
          UCCAGAUGUCACUUGGCAUACACCUGAGUGACUUUAGUGGACAUCUCAAGUUUCCAACUAAGGCACGCGGUGAAUGCCACGUGGCAAUUCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011541
          miRBase ID: crm-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-124a
          Stem-loop Accession # MI0030604
          Stem-loop Sequence
          AUCAAGAUCAGACUCUGCCCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGGCGGCUGCACUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035922
          miRBase ID: chi-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-124-1
          Stem-loop Accession # MI0012662
          Stem-loop Sequence
          CUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012906
          miRBase ID: eca-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID eca-mir-124-2
          Stem-loop Accession # MI0012755
          Stem-loop Sequence
          GUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012906
          miRBase ID: eca-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-124a-1
          Stem-loop Accession # MI0024473
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029396
          miRBase ID: ipu-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-124a-2
          Stem-loop Accession # MI0024472
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029396
          miRBase ID: ipu-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-124a-3
          Stem-loop Accession # MI0024474
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029396
          miRBase ID: ipu-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-124a-4
          Stem-loop Accession # MI0024471
          Stem-loop Sequence
          UGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029396
          miRBase ID: ipu-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID ipu-mir-124a-5
          Stem-loop Accession # MI0024470
          Stem-loop Sequence
          CGUGUUCACGGCGGACCUUGAUUUAAUAUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029396
          miRBase ID: ipu-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-124-1
          Stem-loop Accession # MI0034195
          Stem-loop Sequence
          CGUGUUCACGGCGGACCUUGAUUUAUUAGCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042399
          miRBase ID: mze-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID mze-mir-124-2
          Stem-loop Accession # MI0034201
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042399
          miRBase ID: mze-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID mze-mir-124-3
          Stem-loop Accession # MI0034192
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042399
          miRBase ID: mze-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID mze-mir-124-4
          Stem-loop Accession # MI0034198
          Stem-loop Sequence
          UGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042399
          miRBase ID: mze-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID mze-mir-124-5
          Stem-loop Accession # MI0034202
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042399
          miRBase ID: mze-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-124-1
          Stem-loop Accession # MI0034449
          Stem-loop Sequence
          CGUGUUCACGGCGGACCUUGAUUUAUUAGCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042577
          miRBase ID: nbr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID nbr-mir-124-2
          Stem-loop Accession # MI0034457
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042577
          miRBase ID: nbr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID nbr-mir-124-3
          Stem-loop Accession # MI0034456
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042577
          miRBase ID: nbr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID nbr-mir-124-4
          Stem-loop Accession # MI0034454
          Stem-loop Sequence
          UGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042577
          miRBase ID: nbr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID nbr-mir-124-5
          Stem-loop Accession # MI0034459
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042577
          miRBase ID: nbr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID nbr-mir-124-6
          Stem-loop Accession # MI0034440
          Stem-loop Sequence
          AGUGUUCACAGUGGACCUUGAUUUAAAAUCAGUCAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042577
          miRBase ID: nbr-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-124a-1
          Stem-loop Accession # MI0034702
          Stem-loop Sequence
          AGUGUUCACAGUGGACCUUGAUUUAAAAUCAGUCAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042770
          miRBase ID: oni-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID oni-mir-124a-2
          Stem-loop Accession # MI0034706
          Stem-loop Sequence
          CGUGUUCACGGCGGACCUUGAUUUAUUAGCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042770
          miRBase ID: oni-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID oni-mir-124a-3
          Stem-loop Accession # MI0034707
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042770
          miRBase ID: oni-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID oni-mir-124a-4
          Stem-loop Accession # MI0034710
          Stem-loop Sequence
          UGUGUUCACAGUGGACCUUGAUUUAAUUUCAAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042770
          miRBase ID: oni-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID oni-mir-124a-5
          Stem-loop Accession # MI0034715
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042770
          miRBase ID: oni-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Stem-loop ID oni-mir-124a-6
          Stem-loop Accession # MI0034716
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042770
          miRBase ID: oni-miR-124a
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Polistes canadensis

          Stem-loop ID pca-mir-124
          Stem-loop Accession # MI0037585
          Stem-loop Sequence
          AGAGAGGAUUUGUGAGUCUGUGCUCCUUGCGUUCACUGCGGGCUUCCAUGUGCCAACUUUUCAAAAUUCAUAAGGCACGCGGUGAAUGCCAAGAGCGGACCAAAUUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045864
          miRBase ID: pca-miR-124-3p
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-124-2
          Stem-loop Accession # MI0014816
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015747
          miRBase ID: ppy-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA

          Pristionchus pacificus

          Stem-loop ID ppc-mir-124
          Stem-loop Accession # MI0011133
          Stem-loop Sequence
          GAAGGGUCGUUAAUUCACUUCGCUUUCACCGGUGACUUUAGUGCUUUGUCACAGAGAGAACUAAGGCACGCGGUGAAUGCCAAGAGAAUAACGAAUCCAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011645
          miRBase ID: ppc-miR-124
          Mature miRNA Sequence: UAAGGCACGCGGUGAAUGCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306225_pri
          TaqMan™ Pri-miRNA Assay : Rn03465782_pri
          TaqMan™ Pri-miRNA Assay : Mm03307226_pri
          TaqMan™ Pri-miRNA Assay : Rn03464574_pri
          TaqMan™ Pri-miRNA Assay : Rn03464567_pri
          TaqMan™ Pri-miRNA Assay : Mm03307218_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10691
          Ambion® Pre-miR™ miRNA Precursor : PM10691
          TaqMan™ MicroRNA Assay : 001182
          mirVana® miRNA inhibitor : MH10691
          mirVana® miRNA mimic : MC10691
          TaqMan™ Advanced miRNA Assay : mmu480901_mir
          TaqMan™ Advanced miRNA Assay : 477879_mir


          miRBase Alias:

          Human: hsa-miR-124-3p (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline