Search
Search

| Assay Name: | cel-lin-4 |
|---|---|
| miRBase Accession Number: | MI0000002 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCUGAGACCUCAAGUGUGA |
| Species: | Caenorhabditis elegans, Caenorhabditis brenneri, Caenorhabditis briggsae |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000303, lin-4 |
Mature miRNA Sequence: | UCCCUGAGACCUCAAGUGUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-lin-4-5p | MIMAT0000002 |
| Caenorhabditis brenneri | cbn-lin-4 | MIMAT0029734 |
| Caenorhabditis briggsae | cbr-lin-4 | MIMAT0000464 |