Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478289_mir

          Assay Name: cel-lin-4-5p
          Stem-loop Accession Number: MI0000002
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UCCCUGAGACCUCAAGUGUGA
          Species: Caenorhabditis elegans, Caenorhabditis brenneri, Caenorhabditis briggsae
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478289_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000303, lin-4

          Mature miRNA Sequence:

          UCCCUGAGACCUCAAGUGUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Caenorhabditis elegans cel-lin-4-5p MIMAT0000002
          Caenorhabditis brenneri cbn-lin-4 MIMAT0029734
          Caenorhabditis briggsae cbr-lin-4 MIMAT0000464

          Stem-loop Details



          Caenorhabditis elegans

          Stem-loop ID cel-lin-4
          Stem-loop Accession # MI0000002
          Stem-loop Sequence
          AUGCUUCCGGCCUGUUCCCUGAGACCUCAAGUGUGAGUGUACUAUUGAUGCUUCACACCUGGGCUCUCCGGGUACCAGGACGGUUUGAGCAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000002
          miRBase ID: cel-lin-4-5p
          Mature miRNA Sequence: UCCCUGAGACCUCAAGUGUGA
          Chromosome Location: NA

          Caenorhabditis brenneri

          Stem-loop ID cbn-lin-4-1
          Stem-loop Accession # MI0024907
          Stem-loop Sequence
          UCCCUGAGACCUCAAGUGUGAGCGUACUGUAGAUGCUUCACACCUGGGCUCUCUGGGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029734
          miRBase ID: cbn-lin-4
          Mature miRNA Sequence: UCCCUGAGACCUCAAGUGUGA
          Chromosome Location: NA

          Stem-loop ID cbn-lin-4-2
          Stem-loop Accession # MI0024908
          Stem-loop Sequence
          UCCCUGAGACCUCAAGUGUGAGCGUACUGUAGAUGCUUCACACCUGGGCUCUCUGGGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029734
          miRBase ID: cbn-lin-4
          Mature miRNA Sequence: UCCCUGAGACCUCAAGUGUGA
          Chromosome Location: NA

          Caenorhabditis briggsae

          Stem-loop ID cbr-lin-4
          Stem-loop Accession # MI0000492
          Stem-loop Sequence
          AUCAGAUGCUUUCGGCCUGUUCCCUGAGACCUCAAGUGUGAGCGUUCUGAACAUGCUUCACGCCUGGGCUCUCCGGGUACCAGGACGGUCUGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000464
          miRBase ID: cbr-lin-4
          Mature miRNA Sequence: UCCCUGAGACCUCAAGUGUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000258
          mirVana® miRNA inhibitor : MH10768
          mirVana® miRNA mimic : MC10768

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline