Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000376
          Assay Name: ath-miR399f
          miRBase Accession Number: MI0001025
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 14444997 - 14445114 [+] on Build TAIR10.1
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Species: Arabidopsis, Arabidopsis lyrata, Brachypodium distachyon, Brassica napus, Citrus sinensis, Cucumis melo, Linum usitatissimum, Manihot esculenta, Populus trichocarpa, Ricinus communis, Theobroma cacao, Vitis vinifera
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000376
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000015, MIR399

          Mature miRNA Sequence:

          UGCCAAAGGAGAUUUGCCCGG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR399f MIMAT0000956
          Arabidopsis lyrata aly-miR399a-3p MIMAT0017572
          Arabidopsis lyrata aly-miR399h-3p MIMAT0017672
          Brachypodium distachyon bdi-miR399d MIMAT0035522
          Brassica napus bna-miR399a MIMAT0004449
          Brassica napus bna-miR399b MIMAT0023642
          View More Rows
          Citrus sinensis csi-miR399a-3p MIMAT0018469
          Citrus sinensis csi-miR399e-3p MIMAT0048934
          Cucumis melo cme-miR399c MIMAT0026152
          Linum usitatissimum lus-miR399a MIMAT0027152
          Linum usitatissimum lus-miR399c MIMAT0027154
          Linum usitatissimum lus-miR399e MIMAT0027156
          Manihot esculenta mes-miR399b MIMAT0029280
          Manihot esculenta mes-miR399c MIMAT0029281
          Manihot esculenta mes-miR399d MIMAT0029282
          Manihot esculenta mes-miR399g MIMAT0029285
          Populus trichocarpa ptc-miR399b MIMAT0002045
          Ricinus communis rco-miR399b MIMAT0014197
          Ricinus communis rco-miR399c MIMAT0014198
          Theobroma cacao tcc-miR399b MIMAT0020436
          Theobroma cacao tcc-miR399d MIMAT0020438
          Vitis vinifera vvi-miR399e MIMAT0005730

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR399f
          Stem-loop Accession # MI0001025
          Stem-loop Sequence
          AUAUGCAUUACAGGGCAAGAUCACCAUUGGCAGAGAUCUAUUACUUCAUUCUUGCAUCAUAUGCAUAAAUGUUUGUGGUGAGCUCUCUGCCAAAGGAGAUUUGCCCGGUAAUUCUCUU
          Chromosome Location Chr. 2 - 14444997 - 14445114 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000956
          miRBase ID: ath-miR399f
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: Chr. 2 - 14444997 - 14445114 [+] on Build TAIR10.1

          Arabidopsis lyrata

          Stem-loop ID aly-MIR399a
          Stem-loop Accession # MI0014588
          Stem-loop Sequence
          CAUGUGAGAUGGAUCAAAGGUGGAAAUGUAUUACAGGGUAAGAUCUCUAUUGGCAGGAAACCAUUACUUAGAUCUUUGCAUCUCUUUAUGCAUUGCUUUUAAGUGAGUUCUCUGCCAAAGGAGAUUUGCCCGGUAAUUCUUCUGCACCAAUAUAUUGAUAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017572
          miRBase ID: aly-miR399a-3p
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID aly-MIR399h
          Stem-loop Accession # MI0014637
          Stem-loop Sequence
          AUUACAGGGCAAGAUCUCUAUUGGCAGGAAACCAUUACUUGACCUUUGCAUCUCUUUAUGCAUUGCUUUGUAGUGAGUUCUUGCCAAAGGAGAUUUGCCCGGUAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017672
          miRBase ID: aly-miR399h-3p
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Brachypodium distachyon

          Stem-loop ID bdi-MIR399d
          Stem-loop Accession # MI0029164
          Stem-loop Sequence
          UGCGUGUGAAGGUGCAUUGCAGGGCAACUCCUCCGUUGGCAGAGGGACGACGGAUGGAUGUGGCCGGGGUUCUCCCUUGAAUCCUUGGUCCUGUCCGUCUGCCAAAGGAGAUUUGCCCGGCGAUUCACCACCCACUUCGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035522
          miRBase ID: bdi-miR399d
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Brassica napus

          Stem-loop ID bna-MIR399a
          Stem-loop Accession # MI0005774
          Stem-loop Sequence
          ACAGGGCAAGAUCUCUAUUGGCAGAAAACCAUUACUUAGAUCUUUGCAUCUCUUUCUGCAUUGAUUUGUAGUGAGUUCUCUGCCAAAGGAGAUUUGCCCGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004449
          miRBase ID: bna-miR399a
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID bna-MIR399b
          Stem-loop Accession # MI0020289
          Stem-loop Sequence
          AUUACAGGGCAAGAUCUCUAUUGGCAGGAAACCAUUACUUGACCUUUGCAUCUCUUUAUGCAUUGUUUUGUAGUGAGUUCUCUGCCAAAGGAGAUUUGCCCGGUAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023642
          miRBase ID: bna-miR399b
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Citrus sinensis

          Stem-loop ID csi-MIR399a
          Stem-loop Accession # MI0016712
          Stem-loop Sequence
          CAUUGAGGUGCAUGCAUGCAGUUGCAUAUUACAGGGCAACAUCUCCAUUGGCAGGCUGCAUACUUAUUACAUCCAUCUCAUAUAUACGUAUGUUCUUCAGUCAGUGUGCCUUCUGCCAAAGGAGAUUUGCCCGGCAAUUCCUCUGCAUUUAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018469
          miRBase ID: csi-miR399a-3p
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID csi-MIR399e
          Stem-loop Accession # MI0039669
          Stem-loop Sequence
          CAAAUGAAUCAAGUGUUUAGAUGCAUUACUGGGCAAUUCUCUAUUGGCAGUUGGCUGCAGGGAGGAGCAACCUAGCUCACUGCCAAAGGAGAUUUGCCCGGCAAUUCAUCUGUGCAAUCAUCCUAAUAUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048934
          miRBase ID: csi-miR399e-3p
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Cucumis melo

          Stem-loop ID cme-MIR399c
          Stem-loop Accession # MI0023256
          Stem-loop Sequence
          AUUUCUGGGCAAUUACUCCAUUGGCAGUUGCCACCUUCCUUUUAAAACACACACACACUGACACAUACAUAUAUAUAUAUUAUAUAUUAUGUGAAAUCAUAUUGCCAAAUUUAAGGCAAUAACAAAAUCGCUACUGCCAAAGGAGAUUUGCCCGGUAAUGCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026152
          miRBase ID: cme-miR399c
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Linum usitatissimum

          Stem-loop ID lus-MIR399a
          Stem-loop Accession # MI0021172
          Stem-loop Sequence
          AGAAUGAAUGAGUGUCGGAGUUGAGAAAUCACAGGGCAAGUACCUCUAUUGGCAGGCAGUCACUUAGACAUAUGUACUAAUUUUUCAAUUUGUCUAAGUGCAUUGUCUGCCAAAGGAGAUUUGCCCGGCGAUUCUUCCACUGCUUAUCGAAACAUGUUCCGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0027152
          miRBase ID: lus-miR399a
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID lus-MIR399c
          Stem-loop Accession # MI0021174
          Stem-loop Sequence
          GUACUGUUGCAGUUGGGAAAUUACAGGGCAACUAUCUCUAUUGGCAGGCAGCCAUGCACUUAGAGACAUACUUAACUGUCUAAGUGCACCGUCUGCCAAAGGAGAUUUGCCCGGCGAUUCCUCUACUGCUUAUCGGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0027154
          miRBase ID: lus-miR399c
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID lus-MIR399e
          Stem-loop Accession # MI0021176
          Stem-loop Sequence
          GCAACGAUCUUGUAUGGAUGCAUUACAGGGCAAUUACUCCCUUGGCAGAUAGCUAUAGUACUAGCUAAAUUGAGCAAACAAGUACAGUAAGCUCACUGCCAAAGGAGAUUUGCCCGGCAAUUCAACCCUAUUACGUACUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0027156
          miRBase ID: lus-miR399e
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Manihot esculenta

          Stem-loop ID mes-MIR399b
          Stem-loop Accession # MI0024322
          Stem-loop Sequence
          CAGGGUAAACUCUCCAUUGGCAGGCAGCCACUUACAGAACUAAUUUGCAUUAUUUGUUUUAGUGUGCUCUCUGCCAAAGGAGAUUUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029280
          miRBase ID: mes-miR399b
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID mes-MIR399c
          Stem-loop Accession # MI0024323
          Stem-loop Sequence
          CAGGGUAAACUCUCCAUUGGCAGGCAGCCACUUACAGAACUAAUUUGCAUUGUUUGGUUUAGUGUGCCCUCUGCCAAAGGAGAUUUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029281
          miRBase ID: mes-miR399c
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID mes-MIR399d
          Stem-loop Accession # MI0024324
          Stem-loop Sequence
          CAGGGUAAACUCUCCAUUGGCAGGCAGCCACUUACAGAAUCAAUAUGCAUUAUUUAUUUUAAUCUGCUCUCUGCCAAAGGAGAUUUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029282
          miRBase ID: mes-miR399d
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID mes-MIR399g
          Stem-loop Accession # MI0024327
          Stem-loop Sequence
          CAGGGCAAAACCUCCAUUGGCAUGCAGCCACACUAAGAUUCUCUAUCCUUGUUGUGCAUUCCUAUAGUGUGCUCCAUGCCAAAGGAGAUUUGCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029285
          miRBase ID: mes-miR399g
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Populus trichocarpa

          Stem-loop ID ptc-MIR399b
          Stem-loop Accession # MI0002339
          Stem-loop Sequence
          GGUGCAGUUGCAUUACAGGGCAAGCUUCCAUUGGCAUGCAGCCACUAAGACACGCAAUAUCUAACACUUGAUGCAUUCCUGUAGAGUGCGGCGCCAUGCCAAAGGAGAUUUGCCCGGCAAUUCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002045
          miRBase ID: ptc-miR399b
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Ricinus communis

          Stem-loop ID rco-MIR399b
          Stem-loop Accession # MI0013423
          Stem-loop Sequence
          UUACAGGGCAAAAUCUCCAUUGGCAUGGAACCACUAAGAUAUCCAAACCAAUCAAGCCAGGAUUUCAAUAGUGUGUGCCCUGCCAAAGGAGAUUUGCCCGGCAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014197
          miRBase ID: rco-miR399b
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID rco-MIR399c
          Stem-loop Accession # MI0013424
          Stem-loop Sequence
          UUACAGGGCAAAACCUUCAUUGGCAUGCUGCCAGACUAAGAUGUGGGACGCAUUCCUAUCGUGUGCAUCAUGCCAAAGGAGAUUUGCCCGGCGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014198
          miRBase ID: rco-miR399c
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Theobroma cacao

          Stem-loop ID tcc-MIR399b
          Stem-loop Accession # MI0017520
          Stem-loop Sequence
          UUCAGGGUGCAACUGCAUUACAGGGCAAGAUUUCCAUUGGCAGGCAGCCACUUAUAUGUGCACCCAAACUCAUUCAUUCAUGUGGUGUGCCCUCUGCCAAAGGAGAUUUGCCCGGCAAUUCUUUUGUACUACUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020436
          miRBase ID: tcc-miR399b
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Stem-loop ID tcc-MIR399d
          Stem-loop Accession # MI0017522
          Stem-loop Sequence
          UCUCUGGUGCAGUUGCAUUACAGGGCAAAGAUCUCCAUUGGCAGGCUGCCACUUUGGGUCACGAACCGAAUUAUGUGUUCCUAUAGUGUGUCUUCUGCCAAAGGAGAUUUGCCCGGCAAUUCUUCCGCAUUGAUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020438
          miRBase ID: tcc-miR399d
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA

          Vitis vinifera

          Stem-loop ID vvi-MIR399e
          Stem-loop Accession # MI0006575
          Stem-loop Sequence
          GCAUAUUACAGGGCAAAUUAUCUUUUGGCAGGCAGCCACUUAGAGACACAGCCAAGCCAUGCAUUCUUGUAGUGUGCCCUCUGCCAAAGGAGAUUUGCCCGGCAAUUCUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0005730
          miRBase ID: vvi-miR399e
          Mature miRNA Sequence: UGCCAAAGGAGAUUUGCCCGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          mirVana® miRNA inhibitor : MH10822
          mirVana® miRNA mimic : MC10822


          miRBase Alias:

          Arabidopsis lyrata: aly-miR399a (v18)
          Arabidopsis lyrata: aly-miR399h (v18)
          Arabidopsis lyrata: aly-miR399j (v19)
          Brevundimonas naejangsanensis DSM 23858: bna-miR399 (v18)
          Citrus sinensis: csi-miR399a (v21)
          Oryza sativa: osa-miR399g (v8.1)
          Populus trichocarpa: ptc-miR399c (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline