Search
Search

| Assay Name: | ath-miR399f |
|---|---|
| miRBase Accession Number: | MI0001025 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGCCAAAGGAGAUUUGCCCGG |
| Species: | Arabidopsis, Arabidopsis lyrata, Brachypodium distachyon, Brassica napus, Citrus sinensis, Cucumis melo, Linum usitatissimum, Manihot esculenta, Populus trichocarpa, Ricinus communis, Theobroma cacao, Vitis vinifera |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000015, MIR399 |
Mature miRNA Sequence: | UGCCAAAGGAGAUUUGCCCGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR399f | MIMAT0000956 |
| Arabidopsis lyrata | aly-miR399a-3p | MIMAT0017572 |
| Arabidopsis lyrata | aly-miR399h-3p | MIMAT0017672 |
| Brachypodium distachyon | bdi-miR399d | MIMAT0035522 |
| Brassica napus | bna-miR399a | MIMAT0004449 |
| Brassica napus | bna-miR399b | MIMAT0023642 |
| View More Rows | ||
| Citrus sinensis | csi-miR399a-3p | MIMAT0018469 |
| Citrus sinensis | csi-miR399e-3p | MIMAT0048934 |
| Cucumis melo | cme-miR399c | MIMAT0026152 |
| Linum usitatissimum | lus-miR399a | MIMAT0027152 |
| Linum usitatissimum | lus-miR399c | MIMAT0027154 |
| Linum usitatissimum | lus-miR399e | MIMAT0027156 |
| Manihot esculenta | mes-miR399b | MIMAT0029280 |
| Manihot esculenta | mes-miR399c | MIMAT0029281 |
| Manihot esculenta | mes-miR399d | MIMAT0029282 |
| Manihot esculenta | mes-miR399g | MIMAT0029285 |
| Populus trichocarpa | ptc-miR399b | MIMAT0002045 |
| Ricinus communis | rco-miR399b | MIMAT0014197 |
| Ricinus communis | rco-miR399c | MIMAT0014198 |
| Theobroma cacao | tcc-miR399b | MIMAT0020436 |
| Theobroma cacao | tcc-miR399d | MIMAT0020438 |
| Vitis vinifera | vvi-miR399e | MIMAT0005730 |