Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000435
          Assay Name: hsa-miR-99a
          miRBase Accession Number: MI0000101
          miRBase Version: v22.1
          Chromosome Location: Chr.21: 16539089 - 16539169 [+] on Build GRCh38
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Gadus morhua, Gorilla gorilla, Guinea pig, Horse, Ictalurus punctatus, Lagothrix lagotricha, Macaca nemestrina, Metriaclima zebra, Microcebus murinus, Neolamprologus brichardi, Ophiophagus hannah, Oreochromis niloticus, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pundamilia nyererei, Rabbit, Rhesus monkey, Salmo salar, Tupaia chinensis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000435
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          AACCCGUAGAUCCGAUCUUGUG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-99a-5p MIMAT0000097
          Mouse mmu-miR-99a-5p MIMAT0000131
          Rat rno-miR-99a-5p MIMAT0000820
          Alligator mississippiensis ami-miR-99a-5p MIMAT0038110
          Anolis carolinensis aca-miR-99a-5p MIMAT0021996
          Chicken gga-miR-99a-5p MIMAT0001103
          View More Rows
          Chrysemys picta cpi-miR-99a-5p MIMAT0037710
          Columba livia cli-miR-99-5p MIMAT0038478
          Dasypus novemcinctus dno-miR-99a-5p MIMAT0047594
          Gadus morhua gmo-miR-99-5p MIMAT0044269
          Gorilla gorilla ggo-miR-99a MIMAT0002412
          Guinea pig cpo-miR-99a-5p MIMAT0046941
          Horse eca-miR-99a MIMAT0013184
          Ictalurus punctatus ipu-miR-99a MIMAT0029532
          Lagothrix lagotricha lla-miR-99a MIMAT0002414
          Macaca nemestrina mne-miR-99a MIMAT0002415
          Metriaclima zebra mze-miR-99b MIMAT0042328
          Microcebus murinus mmr-miR-99a MIMAT0049202
          Neolamprologus brichardi nbr-miR-99b MIMAT0042505
          Ophiophagus hannah oha-miR-99a-5p MIMAT0036964
          Oreochromis niloticus oni-miR-99b MIMAT0042695
          Pan paniscus ppa-miR-99a MIMAT0002416
          Pan troglodytes ptr-miR-99a MIMAT0002411
          Pig ssc-miR-99a-5p MIMAT0013896
          Pongo pygmaeus ppy-miR-99a MIMAT0002413
          Pundamilia nyererei pny-miR-99b MIMAT0042888
          Rabbit ocu-miR-99a-5p MIMAT0048182
          Rhesus monkey mml-miR-99a-5p MIMAT0002410
          Salmo salar ssa-miR-99-5p MIMAT0032718
          Tupaia chinensis tch-miR-99a-5p MIMAT0036563
          Xenopus tropicalis xtr-miR-99 MIMAT0003582
          Zebrafish dre-miR-99 MIMAT0001812

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-99a
          Stem-loop Accession # MI0000101
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000097
          miRBase ID: hsa-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-99a
          Stem-loop Accession # MI0000146
          Stem-loop Sequence
          CAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCGCAAGCUCGUUUCUAUGGGUCUGUG
          Chromosome Location Chr. 16 - 77598936 - 77599000 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000131
          miRBase ID: mmu-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: Chr. 16 - 77598936 - 77599000 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-99a
          Stem-loop Accession # MI0000883
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGUUUCUAUGGGUCUGUGGCAGUGUG
          Chromosome Location Chr. 11 - 16052153 - 16052233 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000820
          miRBase ID: rno-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: Chr. 11 - 16052153 - 16052233 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-99a
          Stem-loop Accession # MI0029708
          Stem-loop Sequence
          GUGCCAGUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAAUACACCACACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUGGUUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038110
          miRBase ID: ami-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-99a
          Stem-loop Accession # MI0018894
          Stem-loop Sequence
          GGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUAUAAUGCACAAGCUCGCUUCUAUGGGUCUGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021996
          miRBase ID: aca-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-99a
          Stem-loop Accession # MI0001173
          Stem-loop Sequence
          CCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGUUGAAAUGCACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001103
          miRBase ID: gga-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-99a
          Stem-loop Accession # MI0029444
          Stem-loop Sequence
          UGGUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAGUAUACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCACUCUGGUGAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037710
          miRBase ID: cpi-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-99
          Stem-loop Accession # MI0029945
          Stem-loop Sequence
          GAUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGUUGAAACGCACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUAUGGUGAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038478
          miRBase ID: cli-miR-99-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-99a
          Stem-loop Accession # MI0038954
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047594
          miRBase ID: dno-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-99
          Stem-loop Accession # MI0036171
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCAAAUAUGACAGCACAAGUUCGCUUCUGCAGGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044269
          miRBase ID: gmo-miR-99-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-99a
          Stem-loop Accession # MI0002704
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002412
          miRBase ID: ggo-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-99a
          Stem-loop Accession # MI0038610
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACGGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046941
          miRBase ID: cpo-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-99a-2
          Stem-loop Accession # MI0012929
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCUCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013184
          miRBase ID: eca-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-99a
          Stem-loop Accession # MI0024668
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCGUAAUCGGCAACCCAAGCUCGAUUCUGUGGGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029532
          miRBase ID: ipu-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-99a
          Stem-loop Accession # MI0002706
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002414
          miRBase ID: lla-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-99a
          Stem-loop Accession # MI0002707
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002415
          miRBase ID: mne-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-99b
          Stem-loop Accession # MI0034096
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCGAAUCUGACAGCACAAGCUCGCCUCUGUGGGUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042328
          miRBase ID: mze-miR-99b
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-99a
          Stem-loop Accession # MI0039922
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049202
          miRBase ID: mmr-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-99b
          Stem-loop Accession # MI0034341
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCAAAUGUGACAGCACAAGCUCGCCUCUGUGGGUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042505
          miRBase ID: nbr-miR-99b
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-99a
          Stem-loop Accession # MI0031495
          Stem-loop Sequence
          GAGUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAAUACUGUGCACAAGCUCGUUUCUAUGGGUCAGUGUCAGUCUGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036964
          miRBase ID: oha-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-99b
          Stem-loop Accession # MI0034604
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCAAAUCUGACAGCACAAGCUCGCCUCUGUGGGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042695
          miRBase ID: oni-miR-99b
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-99a
          Stem-loop Accession # MI0002708
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002416
          miRBase ID: ppa-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-99a
          Stem-loop Accession # MI0002703
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002411
          miRBase ID: ptr-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-99a
          Stem-loop Accession # MI0013114
          Stem-loop Sequence
          CCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013896
          miRBase ID: ssc-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-99a
          Stem-loop Accession # MI0002705
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002413
          miRBase ID: ppy-miR-99a
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-99b
          Stem-loop Accession # MI0034850
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCGAAUCUGACAGCACAAGCUCGCCUCUGUGGGUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042888
          miRBase ID: pny-miR-99b
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-99a
          Stem-loop Accession # MI0039264
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048182
          miRBase ID: ocu-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-99a
          Stem-loop Accession # MI0002702
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002410
          miRBase ID: mml-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-99-1
          Stem-loop Accession # MI0026767
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCAAAUCUGACAUCACAAGCUCCUCUCUGUGGGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032718
          miRBase ID: ssa-miR-99-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-99-2
          Stem-loop Accession # MI0026768
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGCGAAUCUGACAUCACAAGCUCGCCUCUGUGGGCCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032718
          miRBase ID: ssa-miR-99-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-99a
          Stem-loop Accession # MI0031223
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036563
          miRBase ID: tch-miR-99a-5p
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-99
          Stem-loop Accession # MI0004821
          Stem-loop Sequence
          UAACAUAAACCCGUAGAUCCGAUCUUGUGGUGAAUUCCUUUGCUCAAGCUCGUUUCUAUGGGUCUGUGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003582
          miRBase ID: xtr-miR-99
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-99-1
          Stem-loop Accession # MI0001956
          Stem-loop Sequence
          GCCACUUGUCAUUAACCCGUAGAUCCGAUCUUGUGAUAAGUUUGAUGGCACAAGCUCGAUUCUAUGGGUCUCUGUCUCUGUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001812
          miRBase ID: dre-miR-99
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA

          Stem-loop ID dre-mir-99-2
          Stem-loop Accession # MI0001957
          Stem-loop Sequence
          CACUUGUCACAAACCCGUAGAUCCGAUCUUGUGGCGUAAUCGGCAACCCAAGCUCGAUUCUGUGGGUCUCUGUCACUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001812
          miRBase ID: dre-miR-99
          Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465023_pri
          TaqMan™ Pri-miRNA Assay : Mm03306206_pri
          TaqMan™ Pri-miRNA Assay : Hs03302729_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10719
          Ambion® Pre-miR™ miRNA Precursor : PM10719
          mirVana® miRNA inhibitor : MH10719
          mirVana® miRNA mimic : MC10719
          TaqMan™ Advanced miRNA Assay : 478519_mir
          TaqMan™ Advanced miRNA Assay : rno478519_mir
          TaqMan™ Advanced miRNA Assay : mmu478519_mir
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          FGMIR : TaqManTM Rodent MicroRNA Array A
          FGMIR : TaqManTM Human MicroRNA Array A
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0


          miRBase Alias:

          Human: hsa-miR-99a (v17)
          Mouse: mmu-miR-99a (v17)
          Rat: rno-miR-99a (v18)
          Aeromonas caviae Ae398: aca-miR-99a (v18)
          Gallus gallus: gga-miR-99a (v18)
          Macaca mulatta: mml-miR-99a (v19)
          Staphylococcus schleiferi: ssc-miR-99a (v21)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline