Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03302729_pri
Assay Name:
hsa-mir-99a
Stem-loop ID:
hsa-mir-99a
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.21: 16539089 - 16539169 [+] on Build GRCh38
Stem-loop Location:
Chr.21: 16539089 - 16539169 [+] on Build GRCh38
Mature miRNA ID:
hsa-miR-99a-3p
hsa-miR-99a-5p
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03302729_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
127
Chromosome Location:
Chr. 21 - 16538684 - 16538684 [-] on 16538684 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-99a-3p
MIMAT0004511
Human
hsa-miR-99a-5p
MIMAT0000097
Stem-loop Details
Human
Stem-loop ID
hsa-mir-99a
Stem-loop Accession #
MI0000101
Stem-loop Sequence
CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
Chromosome Location
Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0004511
miRBase ID: hsa-miR-99a-3p
Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
Chromosome Location: Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38
miRBase Accession #: MIMAT0000097
miRBase ID: hsa-miR-99a-5p
Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG
Chromosome Location: Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38
Gene Family ID
MIPF0000033,
mir-10
Distance to Amplicon
325 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12983
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12983
TaqMan
™
MicroRNA Assay :
002141
mir
Vana
®
miRNA inhibitor :
MH12983
mir
Vana
®
miRNA mimic :
MC12983
TaqMan
™
Advanced miRNA Assay :
479224_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10719
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10719
TaqMan
™
MicroRNA Assay :
000435
mir
Vana
®
miRNA inhibitor :
MH10719
mir
Vana
®
miRNA mimic :
MC10719
TaqMan
™
Advanced miRNA Assay :
478519_mir
TaqMan
™
Advanced miRNA Assay :
rno478519_mir
TaqMan
™
Advanced miRNA Assay :
mmu478519_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline