Search
Search

| Assay Name: | hsa-miR-99a-3p |
|---|---|
| miRBase Accession Number: | MI0000101 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAGCUCGCUUCUAUGGGUCUG |
| Species: | Human, Alligator mississippiensis, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Rabbit |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | CAAGCUCGCUUCUAUGGGUCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-99a-3p | MIMAT0004511 |
| Alligator mississippiensis | ami-miR-99a-3p | MIMAT0038111 |
| Chrysemys picta | cpi-miR-99a-3p | MIMAT0037711 |
| Columba livia | cli-miR-99-3p | MIMAT0038479 |
| Cricetulus griseus | cgr-miR-99a-3p | MIMAT0024017 |
| Dasypus novemcinctus | dno-miR-99a-3p | MIMAT0047595 |
| View More Rows | ||
| Guinea pig | cpo-miR-99a-3p | MIMAT0046942 |
| Rabbit | ocu-miR-99a-3p | MIMAT0048183 |