Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12983
          Assay Name: hsa-miR-99a-3p
          miRBase Accession Number: MI0000101
          miRBase Version: v22.1
          Chromosome Location: Chr.21: 16539089 - 16539169 [+] on Build GRCh38
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Species: Human, Alligator mississippiensis, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Rabbit
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12983

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          CAAGCUCGCUUCUAUGGGUCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-99a-3p MIMAT0004511
          Alligator mississippiensis ami-miR-99a-3p MIMAT0038111
          Chrysemys picta cpi-miR-99a-3p MIMAT0037711
          Columba livia cli-miR-99-3p MIMAT0038479
          Cricetulus griseus cgr-miR-99a-3p MIMAT0024017
          Dasypus novemcinctus dno-miR-99a-3p MIMAT0047595
          View More Rows
          Guinea pig cpo-miR-99a-3p MIMAT0046942
          Rabbit ocu-miR-99a-3p MIMAT0048183

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-99a
          Stem-loop Accession # MI0000101
          Stem-loop Sequence
          CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG
          Chromosome Location Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004511
          miRBase ID: hsa-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-99a
          Stem-loop Accession # MI0029708
          Stem-loop Sequence
          GUGCCAGUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAAUACACCACACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUGGUUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038111
          miRBase ID: ami-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-99a
          Stem-loop Accession # MI0029444
          Stem-loop Sequence
          UGGUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAGUAUACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCACUCUGGUGAUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037711
          miRBase ID: cpi-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-99
          Stem-loop Accession # MI0029945
          Stem-loop Sequence
          GAUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGUUGAAACGCACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUAUGGUGAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038479
          miRBase ID: cli-miR-99-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-99a
          Stem-loop Accession # MI0020564
          Stem-loop Sequence
          CGUUGACAUAAACCCGUAGAUCCGAUCUUGUGGUGAAAGUGGACGCACAAGCUCGCUUCUAUGGGUCUGUGGCAGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024017
          miRBase ID: cgr-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-99a
          Stem-loop Accession # MI0038954
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047595
          miRBase ID: dno-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-99a
          Stem-loop Accession # MI0038610
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACGGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046942
          miRBase ID: cpo-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-99a
          Stem-loop Accession # MI0039264
          Stem-loop Sequence
          AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048183
          miRBase ID: ocu-miR-99a-3p
          Mature miRNA Sequence: CAAGCUCGCUUCUAUGGGUCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302729_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12983
          Ambion® Pre-miR™ miRNA Precursor : PM12983
          TaqMan™ MicroRNA Assay : 002141
          mirVana® miRNA mimic : MC12983
          TaqMan™ Advanced miRNA Assay : 479224_mir


          miRBase Alias:

          Human: hsa-miR-99a* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline