Search
Search

| Assay Name: | hsa-miR-130b |
|---|---|
| miRBase Accession Number: | MI0000748 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUGCAAUGAUGAAAGGGCAU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Xenopus laevis, Xenopus tropicalis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | CAGUGCAAUGAUGAAAGGGCAU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-130b-3p | MIMAT0000691 | |
| Mouse | mmu-miR-130b-3p | MIMAT0000387 | |
| Rat | rno-miR-130b-3p | MIMAT0000837 | |
| Bovine | bta-miR-130b | MIMAT0009224 | |
| Callithrix jacchus | cja-miR-130b | MIMAT0039397 | |
| Cricetulus griseus | cgr-miR-130b-3p | MIMAT0023757 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-130b-3p | MIMAT0047630 | |
| Dog | cfa-miR-130b | MIMAT0006659 | |
| Goat | chi-miR-130b-3p | MIMAT0035946 | |
| Guinea pig | cpo-miR-130b-3p | MIMAT0046983 | |
| Horse | eca-miR-130b | MIMAT0012996 | |
| Pan troglodytes | ptr-miR-130b | MIMAT0008026 | |
| Pig | ssc-miR-130b-3p | MIMAT0013922 | |
| Pongo pygmaeus | ppy-miR-130b | MIMAT0015754 | |
| Rabbit | ocu-miR-130b-3p | MIMAT0048220 | |
| Rhesus monkey | mml-miR-130b-3p | MIMAT0006187 | |
| Xenopus laevis | xla-miR-130b-3p | MIMAT0046444 | |
| Xenopus tropicalis | xtr-miR-130b | MIMAT0003593 | |