Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481434_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-130b-3p
          Stem-loop Accession Number: MI0000748
          miRBase Version: v22.1
          Chromosome Location: Chr.22: 21653304 - 21653385 [+] on Build GRCh38
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Xenopus laevis, Xenopus tropicalis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481434_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          CAGUGCAAUGAUGAAAGGGCAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-130b-3p MIMAT0000691
          Mouse mmu-miR-130b-3p MIMAT0000387
          Rat rno-miR-130b-3p MIMAT0000837
          Bovine bta-miR-130b MIMAT0009224
          Callithrix jacchus cja-miR-130b MIMAT0039397
          Cricetulus griseus cgr-miR-130b-3p MIMAT0023757
          View More Rows
          Dasypus novemcinctus dno-miR-130b-3p MIMAT0047630
          Dog cfa-miR-130b MIMAT0006659
          Goat chi-miR-130b-3p MIMAT0035946
          Guinea pig cpo-miR-130b-3p MIMAT0046983
          Horse eca-miR-130b MIMAT0012996
          Pan troglodytes ptr-miR-130b MIMAT0008026
          Pig ssc-miR-130b-3p MIMAT0013922
          Pongo pygmaeus ppy-miR-130b MIMAT0015754
          Rabbit ocu-miR-130b-3p MIMAT0048220
          Rhesus monkey mml-miR-130b-3p MIMAT0006187
          Xenopus laevis xla-miR-130b-3p MIMAT0046444
          Xenopus tropicalis xtr-miR-130b MIMAT0003593

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-130b
          Stem-loop Accession # MI0000748
          Stem-loop Sequence
          GGCCUGCCCGACACUCUUUCCCUGUUGCACUACUAUAGGCCGCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGUC
          Chromosome Location Chr. 22 - 21653304 - 21653385 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000691
          miRBase ID: hsa-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: Chr. 22 - 21653304 - 21653385 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-130b
          Stem-loop Accession # MI0000408
          Stem-loop Sequence
          GGCUUGUUGGACACUCUUUCCCUGUUGCACUACUGUGGGCCUCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCUGUCGGGCC
          Chromosome Location Chr. 16 - 17124061 - 17124142 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000387
          miRBase ID: mmu-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: Chr. 16 - 17124061 - 17124142 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-130b
          Stem-loop Accession # MI0000904
          Stem-loop Sequence
          GGCUUGCUGGACACUCUUUCCCUGUUGCACUACUGUGGGCCUCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCCGUCAGGCC
          Chromosome Location Chr. 11 - 88129773 - 88129854 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000837
          miRBase ID: rno-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: Chr. 11 - 88129773 - 88129854 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-130b
          Stem-loop Accession # MI0009731
          Stem-loop Sequence
          GGCCUGCCUGACACUCUUUCCCUGUUGCACUACUGUGCGCCCCUGGCAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009224
          miRBase ID: bta-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-130b
          Stem-loop Accession # MI0031916
          Stem-loop Sequence
          UGGCAGGCCUGCCUGAUACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGAAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGUACCGCCUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039397
          miRBase ID: cja-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-130b
          Stem-loop Accession # MI0020395
          Stem-loop Sequence
          CUUGCUGGACACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGGAGCAGUGCAAUGAUGAAAGGGCAUCUGUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023757
          miRBase ID: cgr-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-130b
          Stem-loop Accession # MI0038975
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGCCGCUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047630
          miRBase ID: dno-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-130b
          Stem-loop Accession # MI0008064
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGCCGCUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006659
          miRBase ID: cfa-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-130b
          Stem-loop Accession # MI0030617
          Stem-loop Sequence
          GGCCUGCCUGACACUCUUUCCCUGUUGCACUACUGUGCGCCCCUGGCAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCGGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035946
          miRBase ID: chi-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-130b
          Stem-loop Accession # MI0038634
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGGCUACUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046983
          miRBase ID: cpo-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-130b
          Stem-loop Accession # MI0012748
          Stem-loop Sequence
          GGCCUGCCUGACACUCUUUCCCUGUUGCACUACUGUGGGCUGCUGGAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012996
          miRBase ID: eca-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-130b
          Stem-loop Accession # MI0008528
          Stem-loop Sequence
          GCCUGCCCGACACUCUUUCCCUGUUGCACUACUAUGGGCCGCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008026
          miRBase ID: ptr-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-130b
          Stem-loop Accession # MI0013136
          Stem-loop Sequence
          GCCUGCCUGACACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCAGUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013922
          miRBase ID: ssc-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-130b
          Stem-loop Accession # MI0014823
          Stem-loop Sequence
          GGCCUGCCCGACACUCUUUCCCUGUUGCACUACUAUGGGCCGCUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015754
          miRBase ID: ppy-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-130b
          Stem-loop Accession # MI0039286
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGGACCACUGGGAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048220
          miRBase ID: ocu-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-130b
          Stem-loop Accession # MI0007619
          Stem-loop Sequence
          GGCCUGCCCGACACUCUUUCCCUGUUGCACUACUGUGGGCCACUGGGAAGCAGUGCAAUGAUGAAAGGGCAUCGGUCAGGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006187
          miRBase ID: mml-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-130b
          Stem-loop Accession # MI0038245
          Stem-loop Sequence
          ACUCUUUCCCUGUUGCACUACUGUGCCAGUGAAUAAAGCAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046444
          miRBase ID: xla-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-130b
          Stem-loop Accession # MI0004833
          Stem-loop Sequence
          CCGCCUGACACUCUUUCCCUGUUGCACUACUGUGGCAGUGAAUAAAGCAGUGCAAUGAUGAAAGGGCAUCAGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003593
          miRBase ID: xtr-miR-130b
          Mature miRNA Sequence: CAGUGCAAUGAUGAAAGGGCAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303416_pri
          TaqMan™ Pri-miRNA Assay : Rn03465054_pri
          TaqMan™ Pri-miRNA Assay : Mm03306685_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10777
          Ambion® Pre-miR™ miRNA Precursor : PM10777
          TaqMan™ MicroRNA Assay : 000456
          mirVana® miRNA inhibitor : MH10777
          mirVana® miRNA mimic : MC10777
          TaqMan™ Advanced miRNA Assay : rno481434_mir
          TaqMan™ Advanced miRNA Assay : 477840_mir


          miRBase Alias:

          Staphylococcus schleiferi: ssc-miR-130b (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline