Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000457
          Assay Name: hsa-miR-132
          miRBase Accession Number: MI0000449
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 2049908 - 2050008 [-] on Build GRCh38
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Eptesicus fuscus, Fugu rubripes, Guinea pig, Horse, Ictalurus punctatus, Microcebus murinus, Monodelphis domestica, Otolemur garnettii, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000457
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000065, mir-132

          Mature miRNA Sequence:

          UAACAGUCUACAGCCAUGGUCG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-132-3p MIMAT0000426
          Mouse mmu-miR-132-3p MIMAT0000144
          Rat rno-miR-132-3p MIMAT0000838
          Alligator mississippiensis ami-miR-132-3p MIMAT0038145
          Bovine bta-miR-132 MIMAT0003812
          Callithrix jacchus cja-miR-132 MIMAT0039423
          View More Rows
          Chicken gga-miR-132a-3p MIMAT0049951
          Chrysemys picta cpi-miR-132-3p MIMAT0037749
          Cricetulus griseus cgr-miR-132-3p MIMAT0023759
          Cyprinus carpio ccr-miR-132a MIMAT0026213
          Dasypus novemcinctus dno-miR-132-3p MIMAT0047632
          Eptesicus fuscus efu-miR-132 MIMAT0035002
          Fugu rubripes fru-miR-132 MIMAT0003059
          Guinea pig cpo-miR-132-3p MIMAT0046985
          Horse eca-miR-132 MIMAT0013021
          Ictalurus punctatus ipu-miR-132b MIMAT0029406
          Microcebus murinus mmr-miR-132 MIMAT0049229
          Monodelphis domestica mdo-miR-132-3p MIMAT0004149
          Otolemur garnettii oga-miR-132 MIMAT0049698
          Pig ssc-miR-132 MIMAT0025361
          Pongo pygmaeus ppy-miR-132 MIMAT0015755
          Pteropus alecto pal-miR-132-3p MIMAT0039992
          Rhesus monkey mml-miR-132-3p MIMAT0006188
          Salmo salar ssa-miR-132-3p MIMAT0032332
          Taeniopygia guttata tgu-miR-132 MIMAT0018619
          Tetraodon nigroviridis tni-miR-132 MIMAT0003060
          Xenopus tropicalis xtr-miR-132 MIMAT0003594
          Zebrafish dre-miR-132-3p MIMAT0001829

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-132
          Stem-loop Accession # MI0000449
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC
          Chromosome Location Chr. 17 - 2049908 - 2050008 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000426
          miRBase ID: hsa-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: Chr. 17 - 2049908 - 2050008 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-132
          Stem-loop Accession # MI0000158
          Stem-loop Sequence
          GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCC
          Chromosome Location Chr. 11 - 75173682 - 75173747 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000144
          miRBase ID: mmu-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: Chr. 11 - 75173682 - 75173747 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-132
          Stem-loop Accession # MI0000905
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCUC
          Chromosome Location Chr. 10 - 62014995 - 62015095 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000838
          miRBase ID: rno-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: Chr. 10 - 62014995 - 62015095 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-132
          Stem-loop Accession # MI0029732
          Stem-loop Sequence
          ACGUCGCCAGGGCAACCGUGGCUUUAGAUUGUUACUGUGUGGUGCGUGGGUAACAGUCUACAGCCAUGGUCGCUGGGCCGGACGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038145
          miRBase ID: ami-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-132
          Stem-loop Accession # MI0005028
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003812
          miRBase ID: bta-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-132
          Stem-loop Accession # MI0031942
          Stem-loop Sequence
          GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039423
          miRBase ID: cja-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-132a
          Stem-loop Accession # MI0040692
          Stem-loop Sequence
          ACCGUGGCUUUAGAUUGUUACUGUGCUGCUGGGGGGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049951
          miRBase ID: gga-miR-132a-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-132
          Stem-loop Accession # MI0029471
          Stem-loop Sequence
          UGUCUCCAGGGCAACCGUGGCUUUAGAUUGUUACUGUGGUUGUGUGUUGGUAACAGUCUACAGCCAUGGUCGCUUGGGCUAGACACAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037749
          miRBase ID: cpi-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-132
          Stem-loop Accession # MI0020396
          Stem-loop Sequence
          UCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023759
          miRBase ID: cgr-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-132a
          Stem-loop Accession # MI0023320
          Stem-loop Sequence
          CUUGCUGUCUCCAUGGUGACCGUGGCAUUAGAUUGUUACUGUAGCAACAGCACUUCUGGUAACAGUCUACAGCCAUGGUCGCUAGGGGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026213
          miRBase ID: ccr-miR-132a
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-132
          Stem-loop Accession # MI0038976
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGAAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047632
          miRBase ID: dno-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-132
          Stem-loop Accession # MI0028687
          Stem-loop Sequence
          CUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035002
          miRBase ID: efu-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-132
          Stem-loop Accession # MI0003399
          Stem-loop Sequence
          GUCUCCAUGGUUACCGUGGCAUUAGAUUGUUACUGUAGCAACUGCACCAGUGGUAACAGUCUACAGCCAUGGUCGCUAGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003059
          miRBase ID: fru-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-132
          Stem-loop Accession # MI0038635
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046985
          miRBase ID: cpo-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-132
          Stem-loop Accession # MI0012770
          Stem-loop Sequence
          GGACAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013021
          miRBase ID: eca-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-132b
          Stem-loop Accession # MI0024492
          Stem-loop Sequence
          ACCGUGGCAUUAGAUUGUUACUGUAGGAACAGAAUUUUUGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029406
          miRBase ID: ipu-miR-132b
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-132
          Stem-loop Accession # MI0039957
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049229
          miRBase ID: mmr-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-132
          Stem-loop Accession # MI0005338
          Stem-loop Sequence
          GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCAGGGGUAACAGUCUACAGCCAUGGUCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004149
          miRBase ID: mdo-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-132
          Stem-loop Accession # MI0040482
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049698
          miRBase ID: oga-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-132
          Stem-loop Accession # MI0022129
          Stem-loop Sequence
          CUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025361
          miRBase ID: ssc-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-132
          Stem-loop Accession # MI0014824
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015755
          miRBase ID: ppy-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-132
          Stem-loop Accession # MI0032437
          Stem-loop Sequence
          ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039992
          miRBase ID: pal-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-132
          Stem-loop Accession # MI0007620
          Stem-loop Sequence
          CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006188
          miRBase ID: mml-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-132-1
          Stem-loop Accession # MI0026475
          Stem-loop Sequence
          ACCGUGGCUUUAGAUUGUUACUCUAGAGACAGCACCAUGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032332
          miRBase ID: ssa-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-132-2
          Stem-loop Accession # MI0026476
          Stem-loop Sequence
          ACCGUGGCUUUAGAUUGUUACUGUAGAGACGGUAACAUGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032332
          miRBase ID: ssa-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-132-3
          Stem-loop Accession # MI0026477
          Stem-loop Sequence
          ACCGUGGCAUUAGAUUGUUACUGUAGAAACAGCACCACUGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032332
          miRBase ID: ssa-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-132-4
          Stem-loop Accession # MI0026478
          Stem-loop Sequence
          ACCGUGGCAUUAGAUUGUUACUGUAGCAACAGCACCACUGGUAACAGUCUACAGCCAUGGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032332
          miRBase ID: ssa-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-132
          Stem-loop Accession # MI0016249
          Stem-loop Sequence
          AUCCGACGCCGCCCGGGCGACCGUGGCUUUAGAUAGUUACUGUUCCACUGGGGGAGUAACAGUCUACAGCCAUGGUCGCCGGGACAGGCAGACUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018619
          miRBase ID: tgu-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-132
          Stem-loop Accession # MI0003400
          Stem-loop Sequence
          GUCUCCAUGGUUACCGUGGCAUUAGAUUGUUACUGUAGCAACUGCACCAGCGGUAACAGUCUACAGCCAUGGUCGCUAGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003060
          miRBase ID: tni-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-132
          Stem-loop Accession # MI0004834
          Stem-loop Sequence
          CUGUCUCCAGGGCAACCGUGGCUUUAGAUUGUUACUGUAGUUCUGCAUUGGUAACAGUCUACAGCCAUGGUCGCUCGGGCAAGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003594
          miRBase ID: xtr-miR-132
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-132-1
          Stem-loop Accession # MI0001989
          Stem-loop Sequence
          GUCUCCAUGGCGACCGUGGCAUUAGAUUGUUACUGUAGGAACAGAAUUUUUGGUAACAGUCUACAGCCAUGGUCGCUAGUGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001829
          miRBase ID: dre-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA

          Stem-loop ID dre-mir-132-2
          Stem-loop Accession # MI0001991
          Stem-loop Sequence
          UCCCUUCCUCUGCUGUCUCCAUGCUGACCGUGGCUUUAGAUUGUUACUGUAGCGUUGGCACGUGGUAACAGUCUACAGCCAUGGUCGCCAAGGGGCAGGUGUUACAGCGACUACUGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001829
          miRBase ID: dre-miR-132-3p
          Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465057_pri
          TaqMan™ Pri-miRNA Assay : Mm03306275_pri
          TaqMan™ Pri-miRNA Assay : Hs03303111_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10166
          Ambion® Pre-miR™ miRNA Precursor : PM10166
          mirVana® miRNA inhibitor : MH10166
          mirVana® miRNA mimic : MC10166
          TaqMan™ Advanced miRNA Assay : 477900_mir
          TaqMan™ Advanced miRNA Assay : rno480919_mir
          TaqMan™ Advanced miRNA Assay : mmu480919_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : TaqManTM Human MicroRNA Array A


          miRBase Alias:

          Human: hsa-miR-132 (v17)
          Mouse: mmu-miR-132 (v17)
          Rat: rno-miR-132 (v18)
          Danio rerio: dre-miR-132 (v19)
          Macaca mulatta: mml-miR-132 (v19)
          Monodelphis domestica: mdo-miR-132 (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline