Search
Search

| Assay Name: | hsa-miR-132-3p |
|---|---|
| miRBase Accession Number: | MI0000449 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAACAGUCUACAGCCAUGGUCG |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Eptesicus fuscus, Fugu rubripes, Guinea pig, Horse, Ictalurus punctatus, Microcebus murinus, Monodelphis domestica, Otolemur garnettii, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | UAACAGUCUACAGCCAUGGUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-132-3p | MIMAT0000426 |
| Mouse | mmu-miR-132-3p | MIMAT0000144 |
| Rat | rno-miR-132-3p | MIMAT0000838 |
| Alligator mississippiensis | ami-miR-132-3p | MIMAT0038145 |
| Bovine | bta-miR-132 | MIMAT0003812 |
| Callithrix jacchus | cja-miR-132 | MIMAT0039423 |
| View More Rows | ||
| Chicken | gga-miR-132a-3p | MIMAT0049951 |
| Chrysemys picta | cpi-miR-132-3p | MIMAT0037749 |
| Cricetulus griseus | cgr-miR-132-3p | MIMAT0023759 |
| Cyprinus carpio | ccr-miR-132a | MIMAT0026213 |
| Dasypus novemcinctus | dno-miR-132-3p | MIMAT0047632 |
| Eptesicus fuscus | efu-miR-132 | MIMAT0035002 |
| Fugu rubripes | fru-miR-132 | MIMAT0003059 |
| Guinea pig | cpo-miR-132-3p | MIMAT0046985 |
| Horse | eca-miR-132 | MIMAT0013021 |
| Ictalurus punctatus | ipu-miR-132b | MIMAT0029406 |
| Microcebus murinus | mmr-miR-132 | MIMAT0049229 |
| Monodelphis domestica | mdo-miR-132-3p | MIMAT0004149 |
| Otolemur garnettii | oga-miR-132 | MIMAT0049698 |
| Pig | ssc-miR-132 | MIMAT0025361 |
| Pongo pygmaeus | ppy-miR-132 | MIMAT0015755 |
| Pteropus alecto | pal-miR-132-3p | MIMAT0039992 |
| Rhesus monkey | mml-miR-132-3p | MIMAT0006188 |
| Salmo salar | ssa-miR-132-3p | MIMAT0032332 |
| Taeniopygia guttata | tgu-miR-132 | MIMAT0018619 |
| Tetraodon nigroviridis | tni-miR-132 | MIMAT0003060 |
| Xenopus tropicalis | xtr-miR-132 | MIMAT0003594 |
| Zebrafish | dre-miR-132-3p | MIMAT0001829 |