Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001076
          Assay Name: mmu-miR-424
          miRBase Accession Number: MI0000590
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 53054255 - 53054349 [-] on Build GRCm38
          Mature miRNA Sequence: CAGCAGCAAUUCAUGUUUUGGA
          Species: Mouse, Rat
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001076
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000164, mir-322

          Mature miRNA Sequence:

          CAGCAGCAAUUCAUGUUUUGGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-322-5p MIMAT0000548
          Rat rno-miR-322-5p MIMAT0001619

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-322
          Stem-loop Accession # MI0000590
          Stem-loop Sequence
          CCUCGUUGACUCCGAAGGGCUGCAGCAGCAAUUCAUGUUUUGGAGUAUUGCCAAGGUUCAAAACAUGAAGCGCUGCAACACCCCUUCGUGGGGAA
          Chromosome Location Chr. X - 53054255 - 53054349 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000548
          miRBase ID: mmu-miR-322-5p
          Mature miRNA Sequence: CAGCAGCAAUUCAUGUUUUGGA
          Chromosome Location: Chr. X - 53054255 - 53054349 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-322-1
          Stem-loop Accession # MI0000589
          Stem-loop Sequence
          CCUCGCUGACUCCGAAGGGAUGCAGCAGCAAUUCAUGUUUUGGAGUAUUGCCAAGGUUCAAAACAUGAAGCGCUGCAACACCCCUUCGUGGGAAA
          Chromosome Location Chr. X - 158148161 - 158148255 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001619
          miRBase ID: rno-miR-322-5p
          Mature miRNA Sequence: CAGCAGCAAUUCAUGUUUUGGA
          Chromosome Location: Chr. X - 158584542 - 158584636 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-322-2
          Stem-loop Accession # MI0031763
          Stem-loop Sequence
          CCUCGCUGACUCCGAAGGGAUGCAGCAGCAAUUCAUGUUUUGGAGUAUUGCCAAGGUUCAAAACAUGAAGCGCUGCAACACCCCUUCGUGGGAAA
          Chromosome Location Chr. X - 158584542 - 158584636 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001619
          miRBase ID: rno-miR-322-5p
          Mature miRNA Sequence: CAGCAGCAAUUCAUGUUUUGGA
          Chromosome Location: Chr. X - 158584542 - 158584636 [+] on Build Rnor_6.0


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465418_pri
          TaqMan™ Pri-miRNA Assay : Mm03306922_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11080
          Ambion® Pre-miR™ miRNA Precursor : PM11080
          mirVana® miRNA inhibitor : MH11080
          mirVana® miRNA mimic : MC11080
          TaqMan™ Advanced miRNA Assay : rno481051_mir
          TaqMan™ Advanced miRNA Assay : mmu481051_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : TaqManTM Rodent MicroRNA Array A
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel


          miRBase Alias:

          Mouse: mmu-miR-322 (v17)
          Mouse: mmu-miR-424 (v9.2)
          Rat: rno-miR-424 (v9.2)
          Rat: rno-miR-322 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline