Search
Search

| Assay Name: | mmu-miR-429 |
|---|---|
| miRBase Accession Number: | MI0001642 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAAUACUGUCUGGUAAUGCCGU |
| Species: | Mouse, Rat, Anolis carolinensis, Astatotilapia burtoni, Bovine, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Guinea pig, Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pig, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | UAAUACUGUCUGGUAAUGCCGU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-429-3p | MIMAT0001537 | |
| Rat | rno-miR-429 | MIMAT0001538 | |
| Anolis carolinensis | aca-miR-429-3p | MIMAT0021957 | |
| Astatotilapia burtoni | abu-miR-429b | MIMAT0042172 | |
| Bovine | bta-miR-429 | MIMAT0009315 | |
| Chicken | gga-miR-429-3p | MIMAT0003371 | |
| View More Rows | |||
| Chrysemys picta | cpi-miR-429-3p | MIMAT0037877 | |
| Columba livia | cli-miR-429-3p | MIMAT0038635 | |
| Cricetulus griseus | cgr-miR-429 | MIMAT0023960 | |
| Cyprinus carpio | ccr-miR-429 | MIMAT0026297 | |
| Dasypus novemcinctus | dno-miR-429-3p | MIMAT0047887 | |
| Dog | cfa-miR-429 | MIMAT0001539 | |
| Fugu rubripes | fru-miR-429 | MIMAT0002979 | |
| Gadus morhua | gmo-miR-429-3p | MIMAT0044108 | |
| Guinea pig | cpo-miR-429-3p | MIMAT0047248 | |
| Metriaclima zebra | mze-miR-429b | MIMAT0042347 | |
| Neolamprologus brichardi | nbr-miR-429b | MIMAT0042580 | |
| Oreochromis niloticus | oni-miR-429b | MIMAT0042772 | |
| Ornithorhynchus anatinus | oan-miR-429-3p | MIMAT0007128 | |
| Pig | ssc-miR-429 | MIMAT0020591 | |
| Pteropus alecto | pal-miR-429-3p | MIMAT0040100 | |
| Pundamilia nyererei | pny-miR-429b | MIMAT0042965 | |
| Python bivittatus | pbv-miR-429-3p | MIMAT0039050 | |
| Tetraodon nigroviridis | tni-miR-429 | MIMAT0002980 | |
| Xenopus laevis | xla-miR-429-3p | MIMAT0046602 | |
| Xenopus tropicalis | xtr-miR-429 | MIMAT0003703 | |
| Zebrafish | dre-miR-429a | MIMAT0001624 | |