Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481162_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-429-3p
          Stem-loop Accession Number: MI0001642
          miRBase Version: v22.1
          Chromosome Location: Chr.4: 156053905 - 156053987 [-] on Build GRCm38
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Species: Mouse, Rat, Anolis carolinensis, Astatotilapia burtoni, Bovine, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Guinea pig, Metriaclima zebra, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pig, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481162_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          UAAUACUGUCUGGUAAUGCCGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-429-3p MIMAT0001537
          Rat rno-miR-429 MIMAT0001538
          Anolis carolinensis aca-miR-429-3p MIMAT0021957
          Astatotilapia burtoni abu-miR-429b MIMAT0042172
          Bovine bta-miR-429 MIMAT0009315
          Chicken gga-miR-429-3p MIMAT0003371
          View More Rows
          Chrysemys picta cpi-miR-429-3p MIMAT0037877
          Columba livia cli-miR-429-3p MIMAT0038635
          Cricetulus griseus cgr-miR-429 MIMAT0023960
          Cyprinus carpio ccr-miR-429 MIMAT0026297
          Dasypus novemcinctus dno-miR-429-3p MIMAT0047887
          Dog cfa-miR-429 MIMAT0001539
          Fugu rubripes fru-miR-429 MIMAT0002979
          Gadus morhua gmo-miR-429-3p MIMAT0044108
          Guinea pig cpo-miR-429-3p MIMAT0047248
          Metriaclima zebra mze-miR-429b MIMAT0042347
          Neolamprologus brichardi nbr-miR-429b MIMAT0042580
          Oreochromis niloticus oni-miR-429b MIMAT0042772
          Ornithorhynchus anatinus oan-miR-429-3p MIMAT0007128
          Pig ssc-miR-429 MIMAT0020591
          Pteropus alecto pal-miR-429-3p MIMAT0040100
          Pundamilia nyererei pny-miR-429b MIMAT0042965
          Python bivittatus pbv-miR-429-3p MIMAT0039050
          Tetraodon nigroviridis tni-miR-429 MIMAT0002980
          Xenopus laevis xla-miR-429-3p MIMAT0046602
          Xenopus tropicalis xtr-miR-429 MIMAT0003703
          Zebrafish dre-miR-429a MIMAT0001624

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-429
          Stem-loop Accession # MI0001642
          Stem-loop Sequence
          CCUGCUGAUGGAUGUCUUACCAGACAUGGUUAGAUCUGGAUGCAUCUGUCUAAUACUGUCUGGUAAUGCCGUCCAUCCACGGC
          Chromosome Location Chr. 4 - 156053905 - 156053987 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001537
          miRBase ID: mmu-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: Chr. 4 - 156053905 - 156053987 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-429
          Stem-loop Accession # MI0001643
          Stem-loop Sequence
          UGCCUGCUGAUGGAUGUCUUACCAGACAUGGUUAGAUCUGGAUGUAUCUGUCUAAUACUGUCUGGUAAUGCCGUCCAUCCAUGGC
          Chromosome Location Chr. 5 - 173488331 - 173488415 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001538
          miRBase ID: rno-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: Chr. 5 - 173488331 - 173488415 [-] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-429
          Stem-loop Accession # MI0018866
          Stem-loop Sequence
          AUUCCCACCGCCUGCUGAUCGCUGUCUUACCAGACAAAGUUAGAUCUUAGCUAUCCCCGUCUAAUACUGUCUGGUAAUGCCGUCGAUCGCAUUGGCAAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021957
          miRBase ID: aca-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-429b
          Stem-loop Accession # MI0033903
          Stem-loop Sequence
          GUCUUACCAGACAUGGUUAGAUGUAAUUAUUGGUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042172
          miRBase ID: abu-miR-429b
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-429
          Stem-loop Accession # MI0009829
          Stem-loop Sequence
          AGCCUGCCGAUGGGCGUCUUACCAGACACGGUUAGAUCUGGUCCUUCUGUCUAAUACUGUCUGGUAAUGCCGUCCAUCCCCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009315
          miRBase ID: bta-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-429
          Stem-loop Accession # MI0003714
          Stem-loop Sequence
          GCCUGCUGAUUGCUGUCUUACCAGGCAAAGUUAGAUCUAGCUAUUUCUGUCUAAUACUGUCUGGUAAUGCCGUCAAUCGCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003371
          miRBase ID: gga-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-429
          Stem-loop Accession # MI0029565
          Stem-loop Sequence
          UGCCUGCUGAUUGAUGUCUUACCAGACAAAGUUAGAUCUAGCUAUUUUCGUCUAAUACUGUCUGGUAAUGCCGUCCAUCGCAUUGGCUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037877
          miRBase ID: cpi-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-429
          Stem-loop Accession # MI0030041
          Stem-loop Sequence
          AAGUGCCUGCUGAUUGCUGUCUUACCAGGCAAAGUUAGAUCUAGCUAUUUUUGUCUAAUACUGUCUGGUAAUGCCGUCAAUCGCAUCGGCAAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038635
          miRBase ID: cli-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-429
          Stem-loop Accession # MI0020525
          Stem-loop Sequence
          GGUACACUGCCUGCUGAUGGAUGUCUUACCAGACAUGGUUAGAUCUGGAUGCUCUGUCUAAUACUGUCUGGUAAUGCCGUCCAUCCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023960
          miRBase ID: cgr-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-429
          Stem-loop Accession # MI0023397
          Stem-loop Sequence
          UUGAUGGGCGUCUUACCAGACAUGGUUAGAUCUAAUAAUUUGUGUCUAAUACUGUCUGGUAAUGCCGUCCAUUACAUGCUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026297
          miRBase ID: ccr-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-429
          Stem-loop Accession # MI0039108
          Stem-loop Sequence
          GUCUUACCAGGCAAAGUUAGAUUGCGAUCACUCUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047887
          miRBase ID: dno-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-429
          Stem-loop Accession # MI0001644
          Stem-loop Sequence
          AGCCUGCUGAUGGGCGUCUUACCAGACACGGUUAGAUCUGGGUUCUGGUGUCUAAUACUGUCUGGUAAUGCCGUUCAUCCAUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001539
          miRBase ID: cfa-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-429
          Stem-loop Accession # MI0003301
          Stem-loop Sequence
          CCUGUUGAUAGGCGUCUUACCAGACAUGGUUAGAUGUAAUUAUUGUUGUCUAAUACUGUCUGGUAAUGCCGUCCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002979
          miRBase ID: fru-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-429
          Stem-loop Accession # MI0036046
          Stem-loop Sequence
          GUCUUACCAGACACGGUUAGAUUUAAUGAUCUGUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044108
          miRBase ID: gmo-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-429
          Stem-loop Accession # MI0038772
          Stem-loop Sequence
          GUCUUACCAGACACGGUUAGAUCUGGAUGCUUCUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047248
          miRBase ID: cpo-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-429b
          Stem-loop Accession # MI0034119
          Stem-loop Sequence
          GUCUUACCAGACAUGGUUAGAUGUAAUUAUUGGUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042347
          miRBase ID: mze-miR-429b
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-429b
          Stem-loop Accession # MI0034450
          Stem-loop Sequence
          GUCUUACCAGACAUGGUUAGAUGUAAUUAUUGGUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042580
          miRBase ID: nbr-miR-429b
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-429b
          Stem-loop Accession # MI0034709
          Stem-loop Sequence
          GUCUUACCAGACAUGGUUAGAUGUAAUUAUUGGUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042772
          miRBase ID: oni-miR-429b
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-429
          Stem-loop Accession # MI0006862
          Stem-loop Sequence
          CGGCGUGGAGGUGCCUGCUGAUUGGUGUCUUACCAGACAAAGUUAGAUCUGACUAUUUUCGUCUAAUACUGUCUGGUAAUGCCGUCAAUCACAGCGGCAAGAUCUUCGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007128
          miRBase ID: oan-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-429
          Stem-loop Accession # MI0017991
          Stem-loop Sequence
          GCCGCCGAUGGGUGUCUUACCAGGCACGGUUAGAUCUGGUUCUUCUGUCUAAUACUGUCUGGUAAUGCCGUCCAUUCCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020591
          miRBase ID: ssc-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-429
          Stem-loop Accession # MI0032493
          Stem-loop Sequence
          GUCUUACCAGACACGGUUAGACCUGGGUCCUUCUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040100
          miRBase ID: pal-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-429b
          Stem-loop Accession # MI0034960
          Stem-loop Sequence
          GUCUUACCAGACAUGGUUAGAUGUAAUUAUUGGUGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042965
          miRBase ID: pny-miR-429b
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-429
          Stem-loop Accession # MI0030309
          Stem-loop Sequence
          GCGCCUGCCGAUCGCUGUCUUACCAGACAAAUUUAGAUCUUAAUCUAUUCCUGUCUAAUACUGUCUGGUAAUGCCGUCGAUUCGCAUUGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039050
          miRBase ID: pbv-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-429
          Stem-loop Accession # MI0003302
          Stem-loop Sequence
          AGCCUGUUGAUAGGCGUCUUACCAGACAUGGUUAGAUGUAAUUAUUGUUGUCUAAUACUGUCUGGUAAUGCCGUCCAUUAAAUGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002980
          miRBase ID: tni-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-429-2
          Stem-loop Accession # MI0038334
          Stem-loop Sequence
          GUCUUACCAGACAAGGUUAGAUCUAGUUACUCUCGUCUAAUACUGUCUGGUAAUGCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046602
          miRBase ID: xla-miR-429-3p
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-429
          Stem-loop Accession # MI0004956
          Stem-loop Sequence
          UGCCUGUUGACCAAUGUCUUACCAGACAAGGUUAGAUCUAGUUACUCUCGUCUAAUACUGUCUGGUAAUGCCGUUGGUCACAUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003703
          miRBase ID: xtr-miR-429
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-429a
          Stem-loop Accession # MI0001720
          Stem-loop Sequence
          CUUGUUGAUGGACGUCUUACCAGACAUGGUUAGAUGUAAUAACUUGUGUCUAAUACUGUCUGGUAAUGCCGUCCAUCACAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001624
          miRBase ID: dre-miR-429a
          Mature miRNA Sequence: UAAUACUGUCUGGUAAUGCCGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464825_pri
          TaqMan™ Pri-miRNA Assay : Mm03307499_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10759
          Ambion® Pre-miR™ miRNA Precursor : PM10759
          TaqMan™ MicroRNA Assay : 001077
          mirVana® miRNA inhibitor : MH10759
          mirVana® miRNA mimic : MC10759
          TaqMan™ Advanced miRNA Assay : rno481162_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline