Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001352
          Assay Name: mmu-miR-499
          miRBase Accession Number: MI0003183
          miRBase Version: v22.1
          Chromosome Location: Chr.20: 34990376 - 34990497 [+] on Build GRCh38
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Bovine, Callithrix jacchus, Dog, Goat, Horse, Ictalurus punctatus, Monodelphis domestica, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001352
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000173, mir-499

          Mature miRNA Sequence:

          UUAAGACUUGCAGUGAUGUUU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-499a-5p MIMAT0002870
          Mouse mmu-miR-499-5p MIMAT0003482
          Rat rno-miR-499-5p MIMAT0003381
          Alligator mississippiensis ami-miR-499-5p MIMAT0038301
          Bovine bta-miR-499 MIMAT0003536
          Callithrix jacchus cja-miR-499a MIMAT0039510
          View More Rows
          Dog cfa-miR-499 MIMAT0006655
          Goat chi-miR-499-5p MIMAT0036261
          Horse eca-miR-499-5p MIMAT0013108
          Ictalurus punctatus ipu-miR-499 MIMAT0029521
          Monodelphis domestica mdo-miR-499-5p MIMAT0012753
          Pig ssc-miR-499-5p MIMAT0013877
          Pongo pygmaeus ppy-miR-499-5p MIMAT0015920
          Pteropus alecto pal-miR-499a-5p MIMAT0039820
          Rhesus monkey mml-miR-499-5p MIMAT0006361
          Tupaia chinensis tch-miR-499a-5p MIMAT0036509

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-499a
          Stem-loop Accession # MI0003183
          Stem-loop Sequence
          GCCCUGUCCCCUGUGCCUUGGGCGGGCGGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCCUGGGCAGGGU
          Chromosome Location Chr. 20 - 34990376 - 34990497 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002870
          miRBase ID: hsa-miR-499a-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: Chr. 20 - 34990376 - 34990497 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-499
          Stem-loop Accession # MI0004676
          Stem-loop Sequence
          GGGUGGGCAGCUGUUAAGACUUGCAGUGAUGUUUAGCUCCUCUGCAUGUGAACAUCACAGCAAGUCUGUGCUGCUGCCU
          Chromosome Location Chr. 2 - 155622880 - 155622958 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003482
          miRBase ID: mmu-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: Chr. 2 - 155622880 - 155622958 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-499
          Stem-loop Accession # MI0003721
          Stem-loop Sequence
          GCUGUUAAGACUUGCAGUGAUGUUUAGCUCCUCUCCAUGUGAACAUCACAGCAAGUCUGUGCUGC
          Chromosome Location Chr. 3 - 151138862 - 151138926 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003381
          miRBase ID: rno-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: Chr. 3 - 151138862 - 151138926 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-499
          Stem-loop Accession # MI0029839
          Stem-loop Sequence
          UGAGAGAGCGGCAGUUAAGACUUGCAGUGAUGUUUAGAUAAUGUAUUACAUGAACAUCACUUUAAGUCUGUGCUACUUCUCUCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038301
          miRBase ID: ami-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-499
          Stem-loop Accession # MI0004750
          Stem-loop Sequence
          GGGCGGGCGGCCGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCCACGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003536
          miRBase ID: bta-miR-499
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-499a
          Stem-loop Accession # MI0032037
          Stem-loop Sequence
          GGCCCUGUCCUUCACCCCUGCCCUGUCCCCUGCGCCUUGGGCGGGCGGCCGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCCUGGGCAGGGUUUGGGGGCAAGGGGUGGGAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039510
          miRBase ID: cja-miR-499a
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-499
          Stem-loop Accession # MI0008059
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006655
          miRBase ID: cfa-miR-499
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-499
          Stem-loop Accession # MI0030812
          Stem-loop Sequence
          UCUGUCCCCUGCACCCGGGGCGGGCGGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCCACGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036261
          miRBase ID: chi-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-499
          Stem-loop Accession # MI0012858
          Stem-loop Sequence
          UGUUCUGUCCCCUGCACCCUGGGCGGGCGGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCCAUGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013108
          miRBase ID: eca-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-499-1
          Stem-loop Accession # MI0024643
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAGAGAAAUGUCACAUGAACAUCACUUUAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029521
          miRBase ID: ipu-miR-499
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Stem-loop ID ipu-mir-499-2
          Stem-loop Accession # MI0024642
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAGGUAAUCUGACCCAUGAACAUCACUUUAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029521
          miRBase ID: ipu-miR-499
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-499
          Stem-loop Accession # MI0012510
          Stem-loop Sequence
          GGGCGGGCAGCUGUUAAGACUUGCAGUGAUGUUUAAUUCUUCUCUAUGUGAACAUCACAACAAGUCUAUACAGUUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012753
          miRBase ID: mdo-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-499
          Stem-loop Accession # MI0013096
          Stem-loop Sequence
          UGGGCGGGCGGCCGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013877
          miRBase ID: ssc-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-499
          Stem-loop Accession # MI0014972
          Stem-loop Sequence
          GCCCUGUCCCCUGUGCCUUGGGCGGGCGGCCGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCUUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015920
          miRBase ID: ppy-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-499a
          Stem-loop Accession # MI0032346
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039820
          miRBase ID: pal-miR-499a-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-499
          Stem-loop Accession # MI0007773
          Stem-loop Sequence
          GCCCUGUCCCCGUGUCUUGGGCGGGCAGCUGUUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGCUGCUUCCCGUCCCUACGCUGCCUGGGCAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006361
          miRBase ID: mml-miR-499-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-499a
          Stem-loop Accession # MI0031167
          Stem-loop Sequence
          UUAAGACUUGCAGUGAUGUUUAACUCCUCUCCACGUGAACAUCACAGCAAGUCUGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036509
          miRBase ID: tch-miR-499a-5p
          Mature miRNA Sequence: UUAAGACUUGCAGUGAUGUUU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool A
          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304133_pri
          TaqMan™ Pri-miRNA Assay : Mm03308167_pri
          TaqMan™ Pri-miRNA Assay : Rn03464686_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11352
          Ambion® Pre-miR™ miRNA Precursor : PM11352
          mirVana® miRNA inhibitor : MH11352
          mirVana® miRNA mimic : MC11352
          TaqMan™ Advanced miRNA Assay : rno481402_mir
          TaqMan™ Advanced miRNA Assay : mmu482780_mir
          TaqMan™ Advanced miRNA Assay : 478139_mir
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          FGMIR : TaqManTM Human MicroRNA Array A
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Rodent MicroRNA Array A


          miRBase Alias:

          Human: hsa-miR-499-5p (v17)
          Mouse: mmu-miR-499 (v17)
          Rat: rno-miR-499 (v18)
          Monodelphis domestica: mdo-miR-499 (v19)
          Staphylococcus schleiferi: ssc-miR-499 (v15)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline